Skip to Content
Merck
All Photos(1)

Key Documents

442315

Supelco

2,4-Dimethylaniline

analytical standard

Synonym(s):

1-Amino-2,4-dimethylbenzene, 2,4-Xylidine, 4-Amino-m-xylene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2C6H3NH2
CAS Number:
Molecular Weight:
121.18
Beilstein:
636243
EC Number:
MDL number:
UNSPSC Code:
12000000
PubChem Substance ID:

grade

analytical standard

vapor density

4.2 (vs air)

vapor pressure

0.16 mmHg ( 25 °C)

CofA

current certificate can be downloaded

autoignition temp.

860 °F

expl. lim.

7 %

packaging

ampule of 1000 mg

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.558 (lit.)

bp

218 °C (lit.)

mp

−14.3 °C (lit.)

density

0.98 g/mL at 25 °C (lit.)

application(s)

environmental

format

neat

storage temp.

2-30°C

SMILES string

Cc1ccc(N)c(C)c1

InChI

1S/C8H11N/c1-6-3-4-8(9)7(2)5-6/h3-5H,9H2,1-2H3

InChI key

CZZZABOKJQXEBO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2,4-Dimethylaniline is a dark brown colored liquid, which is obtained as a final degradation product of amitraz, a formamide acaricide.[1][2] It is an environmentally stable compound, which is found to be oncogenic, mutagenic and genotoxic in nature.[2]

Application

2,4-Dimethylaniline can undergo oxidative degradation, to form a series of hydroxylated aromatic amines, in the presence of ultraviolet-visible light or hydrogen peroxide.[3]
2,4-Dimethylaniline may be used as an analytical standard for the determination of the analyte in beeswax, honey, and environmental samples by various chromatography techniques.[4][5][6][7]
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Chronic 2 - STOT RE 2

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

208.4 °F - closed cup

Flash Point(C)

98 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

New evidence against hydroxyl radicals as reactive intermediates in the thermal and photochemically enhanced Fenton reactions
Bossmann.HS, et al.
The Journal of Physical Chemistry A, 102, 5542-5550 (1998)
Voltammetric analysis of the acaricide amitraz and its degradant, 2, 4-dimethylaniline
Brimecombe.R and Limson.J
Talanta, 71, 1298-1303 (2007)
Headspace solvent microextraction-gas chromatographic thermionic specific detector determination of amitraz in honey after hydrolysis to 2, 4-dimethylaniline
Shamsipur M, et al.
Journal of food composition and analysis, 21(3), 264-270 (2008)
Solid phase microextraction and gas chromatography with ion trap detector (GC-ITD) analysis of amitraz residues in beeswax after hydrolysis to 2, 4-dimethylaniline
Lenicek J, et al.
Analytica Chimica Acta, 571(1), 40-44 (2006)
Extraction and clean-up methods for the determination of amitraz total residues in beeswax by gas chromatography with electron capture detection
Jimenez JJ, et al.
Analytica Chimica Acta, 524(1-2), 271-278 (2004)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service