Skip to Content
Merck
All Photos(1)

Key Documents

78113

Supelco

2,4-Dimethylaniline

certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

Synonym(s):

1-Amino-2,4-dimethylbenzene, 2,4-Xylidine, 4-Amino-m-xylene

Sign Into View Organizational & Contract Pricing

Select a Size

78113-100MG
€125.00

€125.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
78113-100MG
€125.00

About This Item

Linear Formula:
(CH3)2C6H3NH2
CAS Number:
Molecular Weight:
121.18
Beilstein:
636243
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

€125.00


Please contact Customer Service for Availability

Request a Bulk Order

grade

certified reference material
TraceCERT®

Quality Level

vapor density

4.2 (vs air)

vapor pressure

0.16 mmHg ( 25 °C)

product line

TraceCERT®

autoignition temp.

860 °F

shelf life

limited shelf life, expiry date on the label

expl. lim.

7 %

manufacturer/tradename

Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

refractive index

n20/D 1.558 (lit.)

bp

218 °C (lit.)

mp

−14.3 °C (lit.)

density

0.98 g/mL at 25 °C (lit.)

format

neat

storage temp.

−20°C

SMILES string

Cc1ccc(N)c(C)c1

InChI

1S/C8H11N/c1-6-3-4-8(9)7(2)5-6/h3-5H,9H2,1-2H3

InChI key

CZZZABOKJQXEBO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI, e.g. NIST or NMIJ.Certified content by quantitative NMR incl. uncertainty and expiry date are given on the certificate. Download your certificate at: http://www.sigma-aldrich.com

Application

2,4-Dimethylaniline is a metabolite of amitraz. It is not listed in the EU pesticide database. The certified reference material (CRM) is intended to be used as a calibrant for chromatography and other analytical techniques.
Environmental

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Chronic 2 - STOT RE 2

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

208.4 °F - closed cup

Flash Point(C)

98 °C - closed cup


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service