Skip to Content
Merck
All Photos(1)

Key Documents

29753-U

Supelco

Carboxen® 569

O.D. × L 1/4 in. × 3 1/2, glass TD tube, fritted, Sealed with Brass Endcaps, preconditioned, pkg of 10 ea

Synonym(s):

Carboxen® 569

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41104008
NACRES:
NB.24

material

glass TD tube

product line

Carboxen®

feature

fritted
preconditioned

packaging

pkg of 10 ea

technique(s)

active air sampling: suitable

O.D. × L

1/4 in. × 3 1/2

matrix

Carboxen® 569 carbon molecular sieve (CMS)

application(s)

air monitoring
environmental
industrial hygiene

compatibility

for sampling (For Sampling in high humidity conditions)
for use with PerkinElmer, Markes, DANI, OI Analytical, and Shimadzu Instruments

Looking for similar products? Visit Product Comparison Guide

Legal Information

Carboxen is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Filippo Maggi et al.
Chemistry & biodiversity, 8(1), 95-114 (2011-01-25)
Headspace solid-phase microextraction (HS-SPME) coupled with GC/FID and GC/MS was applied for the first time in the analysis of the volatile fraction of an Ephedra species. Notably, six Italian populations (Marche, Abruzzo, and Sardinia) of Ephedra nebrodensis subsp. nebrodensis, covering
Jacek A Koziel et al.
Journal of the Air & Waste Management Association (1995), 55(8), 1147-1157 (2005-09-29)
Odorous gases associated with livestock operations are complex mixtures of hundreds if not thousands of compounds. Research is needed to know how best to sample and analyze these compounds. The main objective of this research was to compare recoveries of
Frank Sporkert et al.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences, 772(1), 45-51 (2002-05-23)
A new and in part automated headspace solid-phase microextraction method for quantitative determination of the highly toxic rodenticide fluoroacetic acid (FAA) in serum and other biological samples has been developed. FAA and deuterated acetic acid (internal standard) were extracted from
Louise Urruty et al.
Journal of agricultural and food chemistry, 50(11), 3129-3136 (2002-05-16)
To provide an efficient and running analytical tool to strawberry plant breeders who have to characterize and compare the aromatic properties of new cultivars to those already known, a HS-SPME/GC-MS analysis method has been coupled with a statistical treatment method
H C Shih et al.
Journal of occupational and environmental hygiene, 9(7), 427-436 (2012-06-02)
A solid-phase microextraction (SPME) device was used as a diffusive sampler for airborne propylene glycol ethers (PGEs), including propylene glycol monomethyl ether (PGME), propylene glycol monomethyl ether acetate (PGMEA), and dipropylene glycol monomethyl ether (DPGME). Carboxen-polydimethylsiloxane (CAR/PDMS) SPME fiber was

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service