Merck
  • Time-weighted average sampling of airborne propylene glycol ethers by a solid-phase microextraction device.
Time-weighted average sampling of airborne propylene glycol ethers by a solid-phase microextraction device.
Journal of occupational and environmental hygiene (2012-06-02)
H C Shih, S W Tsai, C H Kuo
ABSTRACT
A solid-phase microextraction (SPME) device was used as a diffusive sampler for airborne propylene glycol ethers (PGEs), including propylene glycol monomethyl ether (PGME), propylene glycol monomethyl ether acetate (PGMEA), and dipropylene glycol monomethyl ether (DPGME). Carboxen-polydimethylsiloxane (CAR/PDMS) SPME fiber was selected for this study. A polytetrafluoroethylene (PTFE) tubing was used as the holder, and the SPME fiber assembly was inserted into the tubing as a diffusive sampler. The diffusion path length and area of the sampler were 0.3 cm and 0.00086 cm(2), respectively. The theoretical sampling constants at 30°C and 1 atm for PGME, PGMEA, and DPGME were 1.50 × 10(-2), 1.23 × 10(-2) and 1.14 × 10(-2) cm(3) min(-1), respectively. For evaluations, known concentrations of PGEs around the threshold limit values/time-weighted average with specific relative humidities (10% and 80%) were generated both by the air bag method and the dynamic generation system, while 15, 30, 60, 120, and 240 min were selected as the time periods for vapor exposures. Comparisons of the SPME diffusive sampling method to Occupational Safety and Health Administration (OSHA) organic Method 99 were performed side-by-side in an exposure chamber at 30°C for PGME. A gas chromatography/flame ionization detector (GC/FID) was used for sample analysis. The experimental sampling constants of the sampler at 30°C were (6.93 ± 0.12) × 10(-1), (4.72 ± 0.03) × 10(-1), and (3.29 ± 0.20) × 10(-1) cm(3) min(-1) for PGME, PGMEA, and DPGME, respectively. The adsorption of chemicals on the stainless steel needle of the SPME fiber was suspected to be one of the reasons why significant differences between theoretical and experimental sampling rates were observed. Correlations between the results for PGME from both SPME device and OSHA organic Method 99 were linear (r = 0.9984) and consistent (slope = 0.97 ± 0.03). Face velocity (0-0.18 m/s) also proved to have no effects on the sampler. However, the effects of temperature and humidity have been observed. Therefore, adjustments of experimental sampling constants at different environmental conditions will be necessary.
Product Description
Supelco
Carboxen® 569, glass TD tube, fritted, O.D. × L 1/4 in. × 3 1/2 in., preconditioned, pkg of 10 ea
Supelco
Tenax® TA / Carboxen® 1018, glass TD tube, preconditioned, O.D. × L 1/4 in. × 3 1/2 in., Sealed with (Swagelok® End-Fittings), pkg of 10 ea
Supelco
ORBO 78 HBr on Carboxen® 564 specially cleaned (20/45), 400/200 mg, W,W,W separators, O.D. × L 6 mm × 110 mm, pkg of 25 ea
Supelco
ORBO 90 Carboxen® 564 (20/45), 160/80 mg, W,F,F separators, O.D. × L 6 mm × 75 mm, pkg of 25 ea, for analyte group MEK (methylethyl ketone)
Sigma-Aldrich
1-Methoxy-2-propanol, ≥99.5%
Supelco
SPME fiber assembly Carboxen/Polydimethylsiloxane (CAR/PDMS), df 75 μm(CAR/PDMS, needle size 23 ga, for use with autosampler
Supelco
SPME fiber assembly Carboxen/Polydimethylsiloxane (CAR/PDMS), df 85 μm(CAR/PDMS, needle size 24 ga, StableFlex, for use with autosampler
Supelco
Carboxen®-1010 PLOT Capillary GC Column, L × I.D. 30 m × 0.53 mm, average thickness 30 μm
Supelco
Carboxen® Adsorbent, matrix Carboxen® 564, 20-45 mesh, pack of 144 × 290 mg
Supelco
Carboxen®-1010 PLOT Capillary GC Column, L × I.D. 30 m × 0.32 mm, average thickness 15 μm
Supelco
Carboxen® Adsorbent, matrix Carboxen® 1003, 40-60 mesh, bottle of 10 g
Supelco
Carboxen® Adsorbent, matrix Carboxen® 1000, 60-80 mesh, bottle of 10 g
Supelco
SPME fiber assembly Carboxen/Polydimethylsiloxane (CAR/PDMS), df 75 μm(CAR/PDMS, for use with manual holder, needle size 24 ga
Supelco
SPME fiber assembly Carboxen/Polydimethylsiloxane (CAR/PDMS), df 75 μm(CAR/PDMS, needle size 24 ga, for use with autosampler
Supelco
Carboxen®-1006 PLOT Capillary GC Column, L × I.D. 30 m × 0.53 mm, average thickness 30 μm
Supelco
Carboxen® Adsorbent, matrix Carboxen® 569, 20-45 mesh, bottle of 10 g
Sigma-Aldrich
Di(propylene glycol) methyl ether, mixture of isomers, 97%
Supelco
1-Methoxy-2-propanol, analytical standard
Supelco
Carboxen® Adsorbent, matrix Carboxen® 572, 20-45 mesh, bottle of 10 g
Supelco
SPME Fiber Assembly, 50/30µm DVB/CAR/PDMS, Metal Alloy (1cm), needle size 23 ga, Autosampler, pk of 1 ×, plain gray hub
Supelco
Carboxen® 1000 Reversible SPE Tube, 200 mg/mL, pk 30
Supelco
SPME Fiber Assembly, 50/30µm DVB/CAR/PDMS, StableFlex (1cm), needle size 24 ga, Manual Holder, pk of 3, plain gray hub
Supelco
Carboxen® Adsorbent, matrix Carboxen® 1000, 40-60 mesh, bottle of 50 g
Supelco
SPME Fiber Assembly, 50/30µm DVB/CAR/PDMS, StableFlex (1 cm), needle size 23 ga, Autosampler, pk of 3, plain gray hub
Supelco
Carboxen® Adsorbent, matrix Carboxen® 569, 20-45 mesh, bottle of 500 g
Supelco
Carboxen® 572 for Tobacco Smoke, Adsorbent Bed Wt: 300 mg, cartridge size 3 mL, pkg of 50 ea
Supelco
ORBO-93 Sorbent Tube, ORBO tube I.D. × L 6 mm × 95 mm, Bed A 180 mg, Bed B 90 mg, 60/80 mesh, pkg of 25 ea
Supelco
SPME fiber assembly Carboxen/Polydimethylsiloxane (CAR/PDMS), df 85 μm(CAR/PDMS, for use with autosampler, needle size 23 ga, metal alloy fiber
Supelco
SPME Fiber Assembly, 50/30µm DVB/CAR/PDMS, Metal Alloy (2cm), needle size 23 ga, Autosampler, pk of 1 ×, plain gray hub
Sigma-Aldrich
Di(propylene glycol) methyl ether, mixture of isomers, ≥99%