Skip to Content
Merck
All Photos(1)

Key Documents

24920-U

Supelco

Purge/Trap K

Vocarb® 3000, for use with StableFlex 3000, 3100 (Straight, 12 in./30.5 cm, Valco fitting

Synonym(s):

PURGE TRAP K, Tekmar® 3000

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
€119.00

€119.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
1 EA
€119.00

About This Item

UNSPSC Code:
12352200
NACRES:
NB.21

€119.00


Please contact Customer Service for Availability

Request a Bulk Order

Product Name

Purge/Trap K Vocarb® 3000, for use with Tekmar® 3000, 3100 (Straight, 12 in./30.5 cm, Valco fitting

Agency

suitable for EPA 601

Quality Level

application(s)

environmental

compatibility

for use with Tekmar® 3000, 3100 (Straight, 12 in./30.5 cm, Valco fitting

Looking for similar products? Visit Product Comparison Guide

General description

VOCARB purge traps efficiently trap analytes in US EPA Methods 502.2 and 524.

Legal Information

Tekmar is a registered trademark of Tekmar Co.
Vocarb is a registered trademark of Siemens Industry, Inc.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service