Skip to Content
Merck
All Photos(1)

Key Documents

20080-U

Supelco

Purge/Trap K

Vocarb® 3000, for use with Encon

Synonym(s):

VOCARB purge trap (Encon)

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
€103.00

€103.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
1 EA
€103.00

About This Item

UNSPSC Code:
23151817
NACRES:
NB.21

€103.00


Please contact Customer Service for Availability

Request a Bulk Order

Product Name

Purge/Trap K Vocarb® 3000, for use with Encon

manufacturer/tradename

Purge Trap K

Quality Level

application(s)

environmental

compatibility

for use with Encon

Looking for similar products? Visit Product Comparison Guide

General description

Purge/Traps K Vocarb® 3000 (for use with Encon) efficiently trap analytes in US EPA Methods 502.2 and 524.

Legal Information

Vocarb is a registered trademark of Siemens Industry, Inc.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service