Key Documents
23779
Supelco
Capillary Column Tags
pkg of 100 ea
Select a Size
100 EA
€30.20
€30.20
Please contact Customer Service for Availability
Request a Bulk Order
Select a Size
Change View
100 EA
€30.20
About This Item
UNSPSC Code:
41115700
€30.20
Please contact Customer Service for Availability
Request a Bulk Order
material
0.016″ aluminum
packaging
pkg of 100 ea
tongue L
5 1/4 in.
writing surface L × W
1 1/8 in. × 3/4 in.
General description
Permanently label capillary columns.
  • Soft enough to imprint with a ball-point pen
  • Big enough for all necessary information
  • Lightweight and will not damage column
Choose from one of the most recent versions:
Certificates of Analysis (COA)