Skip to Content
Merck
All Photos(1)

Documents

20421

Supelco

GR-2 Septa

diam. 11 mm ( 7/16 in.)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121800

packaging

pkg of 100 ea

technique(s)

gas chromatography (GC): suitable

diam.

11 mm ( 7/16 in.)

Looking for similar products? Visit Product Comparison Guide

General description

These low cost gray silicone rubber septa are designed for routine, isothermal use. For use with 50 °C to 200 °C inlet temperature.

Application

GR-2 silicon rubber septa maybe fitted to make a gas-tight seal between the glass opening and the steel gas fitting.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Susan Wagner et al.
Molecular cell, 79(4), 546-560 (2020-06-27)
Translational control targeting the initiation phase is central to the regulation of gene expression. Understanding all of its aspects requires substantial technological advancements. Here we modified yeast translation complex profile sequencing (TCP-seq), related to ribosome profiling, and adapted it for
H2 production from marine and freshwater species of green algae during sulfur deprivation and considerations for bioreactor design.
Kari S.
International Journal of Hydrogen Energy, 33 (2), 511-521 (2008)
Megan J Agajanian et al.
Cell reports, 26(1), 79-93 (2019-01-04)
β-Catenin-dependent WNT signal transduction governs development, tissue homeostasis, and a vast array of human diseases. Signal propagation through a WNT-Frizzled/LRP receptor complex requires proteins necessary for clathrin-mediated endocytosis (CME). Paradoxically, CME also negatively regulates WNT signaling through internalization and degradation
Sayantani Sinha et al.
Cell reports, 31(4), 107570-107570 (2020-04-30)
Bone morphogenic protein (BMP)/transforming growth factor β (TGF-β) signaling determines mesenchymal-stromal-cell (MSC) osteolineage commitment and tissue identity. However, molecular integration of developmental signaling with MSC-intrinsic chromatin regulation remains incompletely understood. SWI/SNF-(BAF) is an ATP-dependent chromatin remodeler implicated in multi-cellular development.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service