Skip to Content
Merck
All Photos(1)

Key Documents

1.11117

Supelco

Ammonium Test Kit, colorimetric

0.5-10 mg/L (NH₄⁺), MQuant®

Sign Into View Organizational & Contract Pricing

Select a Size

150 TESTS
€93.30

€93.30


Estimated to ship on09 May 2025Details


Request a Bulk OrderRequest more information

Select a Size

Change View
150 TESTS
€93.30

About This Item

UNSPSC Code:
41116116
NACRES:
NA.24

€93.30


Estimated to ship on09 May 2025Details


Request a Bulk OrderRequest more information

Product Name

Ammonium Test, colorimetric, 0.5-10 mg/L (NH4+), for use with MQUANT®

specific analyte(s)

ammonium

Quality Level

measuring range

0.5-10 mg/L (NH4+)

compatibility

for use with MQUANT®

detection method

colorimetric

storage temp.

15-25°C

General description

Method: colorimetric, acc. to Neßler, with color card and sliding comparator 0.5 - 1 - 2 - 3 - 5 - 7 - 10 mg/l NH₄⁺ MQuant®
This MQuant® Ammonium test (formally MColortest®) is suitable for the colorimetric determination using the easy to use color card. The evaluation considers the untreated sample so even turbid and colored water samples can be analyzed. The test measures both ammonium ions and dissolved ammonia in e.g. groundwater and surface water, seawater, drinking water and aquarium water. Ammonium nitrogen (NH4-N) occurs partly in the form of ammonium ions and partly as ammonia. A pH-dependent equilibrium exists between the two forms.

Method
Ammonium ions react with Neßler′s reagent to form a yellow-brown compound. The ammonium concentration is measured semiquantitatively by allocating the color to a value on a reference scale.

The test can be used in:
  • Groundwater and surface water
  • Drinking water
  • Wastewater
  • Aquarium water, waters from aquaculture
  • Boiler and boiler feed water, cooling water
  • Swimming-pool water
  • Soils and fertilizers after appropriate sample pretreatment

This test is not suited for seawater.

Legal Information

MCOLORTEST is a registered trademark of Merck KGaA, Darmstadt, Germany
MQUANT is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

For U.S. Customers: Contains mercury; Do not place in trash - dispose according to local, state, or federal laws.

Signal Word

Danger

Hazard Classifications

Acute Tox. 1 Dermal - Acute Tox. 2 Inhalation - Acute Tox. 2 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1A - STOT RE 2

Storage Class Code

6.1B - Non-combustible, acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service