Skip to Content
Merck
All Photos(1)

Key Documents

11501-U

Supelco

PTFE Packed GC Column (General Configuration)

phase none, matrix Chromosil 310 support, L × O.D. × I.D. 8.0 ft (2.4 m) × 1/8 in. × 2.1 mm, Ends empty; middle 6 ft packed

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
€399.00

€399.00


Please contact Customer Service for Availability


Select a Size

Change View
1 EA
€399.00

About This Item

UNSPSC Code:
41115710
NACRES:
SB.54

€399.00


Please contact Customer Service for Availability

material

PTFE

technique(s)

gas chromatography (GC): suitable

L × O.D. × I.D.

8.0 ft (2.4 m) × 1/8 in. × 2.1 mm, Ends empty; middle 6 ft packed

matrix

Chromosil 310 support

matrix active group

none phase

application(s)

pharmaceutical (small molecule)

column type

packed GC

compatibility

configured for general coil use
for analyte group used with sulfur compounds
for use with General Configuration

Looking for similar products? Visit Product Comparison Guide

General description

For low temperature applications, a PTFE column offers the flexibility of metal with inertness approaching that of glass. This column is of a general configuration, and can be carefully bent to fit most instruments.

Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Met. Corr. 1

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

  1. How is compatibility with Chromosil 310 and BF3? I'd like to use it for analyzing SO2 in BF3.

    1 answer
    1. Boron trifluoride, a strong Lewis acid, can react with siloxane-based materials, potentially causing degradation or alteration of the stationary phase. Therefore, using Chromosil 310 with boron trifluoride may not be ideal, as it can lead to compromised column performance, including changes in retention times, peak shapes, or even complete column failure.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service