Skip to Content
Merck
All Photos(11)

Key Documents

ZRB1646

Sigma-Aldrich

Anti-SLC16A3/MCT4 Antibody, clone 2J22 ZooMAb® Rabbit Monoclonal

enhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Synonym(s):

Monocarboxylate transporter 4;MCT 4;Solute carrier family 16 member 3

Sign Into View Organizational & Contract Pricing

Select a Size

25 μL
€191.00
4 X 25 μL
€393.00

€191.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
25 μL
€191.00
4 X 25 μL
€393.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

€191.00


Please contact Customer Service for Availability

Request a Bulk Order

biological source

rabbit

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

2J22, recombinant monoclonal

description

recombinant, expressed in HEK 293 cells

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt 49.47 kDa
observed mol wt ~55 kDa

purified by

using Protein A

species reactivity

human

packaging

antibody small pack of 25 μL

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

sustainability

Greener Alternative Product

technique(s)

affinity binding assay: suitable
immunocytochemistry: suitable
immunohistochemistry: suitable
western blot: suitable

isotype

IgG

epitope sequence

C-terminal

Protein ID accession no.

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

human ... SLC16A3(9123)

General description

ZooMAb® antibodies represent an entirely new generation of recombinant monoclonal antibodies.Each ZooMAb® antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb® antibodies are reliably available and ready to ship when you need them.

Specificity

Clone 2J22 is a ZooMAb® Rabbit recombinant monoclonal antibody that specifically detects human Monocarboxylate transporter 4 (SLC16A3). It targets an epitope within 22 amino acids from the C-terminal, cytoplasmic domain.

Immunogen

KLH-conjugated linear peptide corresponding to 22 amino acids from the C-terminal, cytoplasmic domain of human Monocarboxylate transporter 4 (SLC16A3).

Application

Quality Control Testing

Evaluated by Western Blotting in HeLa cell lysate.

Western Blotting Analysis: A 1:1,000 dilution of this antibody detected SLC16A3/MCT4 in HeLa cell lysate.

Tested Applications

Western Blotting Analysis: A 1:1,000 dilution from a representative lot detected SLC16A3/MCT4 in A431 cell lysate.

Affinity Binding Assay: A representative lot of this antibody bound SLC16A3/MCT4 peptide with a KD of 3.5 x 10-7 in an affinity binding assay.

Immunocytochemistry Analysis: A 1:100 dilution from a representative lot detected SLC16A3/MCT4 in A431 cells.

Immunohistochemistry (Paraffin) Analysis: A 1:100 dilution from a representative lot detected SLC16A3/MCT4 in human skeletal muscle tissue sections.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user.

Target description

Monocarboxylate transporter 4 (UniProt: O15427; also known as MCT 4, Solute carrier family 16 member 3) is encoded by the SLC16A3 (also known as MCT3, MCT4) gene (Gene ID: 9123) in human. MCT4 is a multi-pass membrane protein with seven cytoplasmic domains, twelve transmembrane domains, and six extracellular domains with amino and carboxyl termini facing the cytosol. It contains two basolateral sorting signals (aa 423-441; 441-465) in its C-terminal cytoplasmic tail that are essential to direct MCT4 to the basolateral membrane. It is the major monocarboxylate transporter present in skeletal muscle that mediates the transport of lactate, pyruvate, and ketone bodies across the plasma membrane. It is also detected in tumor cells and white blood cells that have high glycolytic rate. It displays higher transport efficiency for L-lactate when compared to D-lactate and pyruvate (Km = 28, 513, and 153 mM, respectively in Xenopus oocytes). Human MCT contains six conserved arginine residues and arginine 278 is reported to be the crucial residue involved in L-lactate recognition. Any mutation in this residue is shown to void L-lactate transport. High levels of MCT4 are reported in head and neck squamous cell carcinoma (HNSCC) and these higher levels are associated with poor prognosis. It is not expressed in normal oral mucosa but is detected in the transformed epithelium. This ZooMAb® recombinant monoclonal antibody, generated by our propriety technology, offers significantly enhanced specificity, affinity, reproducibility, and stability over conventional monoclonals. (Ref.: Bisetto, S., et al (2018). Front. Oncol. 8; 324; Sasaki, S., et al. (2013). PLoS One.8(7); e67690; Manning Fox, JE., et al. (2000). J. Physiol. 529(Pt2); 285-293).

Physical form

Purified recombinant rabbit monoclonal antibody IgG, lyophilized in PBS, 5% Trehalose, normal appearance a coarse or translucent resin. The PBS/trehalose components in the ZooMAb formulation can have the appearance of a semi-solid (bead like gel) after lyophilization. This is a normal phenomenon. Please follow the recommended reconstitution procedure in the data sheet to dissolve the semi-solid, bead-like, gel-appearing material. The resulting antibody solution is completely stable and functional as proven by full functional testing. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 µL.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 µL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service