Z744262
Wilmad® NMR Benchtop Tube
tube O.D. 5 mm, tube L 8 in., pack of 150
Synonym(s):
NMR tube, spectrometer NMR tube
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
packaging
pack of 150
manufacturer/tradename
Wilmad WG-BTNMR-8
tube L
8 in.
tube O.D.
5 mm
suitability
suitable for NMR
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Ideal for use with 43, 60,& 80MHz manual sample loading benchtop NMR spectrometers, NMR Tubes have been tested in the most popular benchtop spectrometers to assure performance.
Type 1, Class B Borosilicate glass construction with disposable caps
Note: Not for use with high-field instruments or spinning experiments
Type 1, Class B Borosilicate glass construction with disposable caps
Note: Not for use with high-field instruments or spinning experiments
Application
NMR
Features and Benefits
Packaging allows for easy tube access and storage
Legal Information
Wilmad is a registered trademark of SP Industries
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service