Yes, both Products Z740301 and Z740302 are individually wrapped
Z740302
Disposable Plastic BioSafe Pipette
capacity 10 mL
Synonym(s):
barrier filter, barrier filter pipette, pipet, pipette, plugged pipette, serological, serum
Select a Size
€105.00
Select a Size
About This Item
€105.00
Recommended Products
material
plastic (Disposable)
sterility
non-sterile
feature
graduations Subdivisions 0.1mL
plugged
packaging
pack of 200 ea
manufacturer/tradename
Cole Parmer 1300253
graduations and accuracy measuring range
0.1 mL
capacity
10 mL
Looking for similar products? Visit Product Comparison Guide
General description
The first pipette with total contamination control
The Argos BioSafe pipette is a standard serological pipette with a unique barrier filter. If over-aspiration occurs with BioSafe pipettes, liquid is “locked” and unable to pass through to filter. Similar in theory to a barrier pipette tip, BioSafe pipettes provide sample integrity while promoting user safety. It also protects electronic pipettors from contamination and cross contaminating samples by over aspiration.
Ideal in applications involving any potentially hazardous or dangerous biological fluids, infectious diseases or when cross over contamination is an issue.
- Unique filter plug “locks” liquid in pipette during over-aspiration
- Available in 5, 10 & 25 mL (individually wrapped)
- Eliminate aerosol or liquid contamination
- Total contamination protection for pipettor and samples
- Use in place of filtered pipette tips
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
-
Are the products individually wrapped?
1 answer-
Helpful?
-
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service