Skip to Content
Merck
All Photos(1)

Key Documents

Z720461

Nunc® EZ Flip conical centrifuge tubes

15 mL conical tube, bulk packed

Synonym(s):

centrifuge tubes

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121703
NACRES:
NB.11

material

cap (leakproof cap)
conical bottom tube
polypropylene

sterility

sterile

feature

RCF 8,500 × g

packaging

pack of 50
case of 500

manufacturer/tradename

Nunc 362694

capacity

15 mL

color

clear

Looking for similar products? Visit Product Comparison Guide

General description

EZ Flip conical centrifuge tubes
Ergonomic design has a hinged-cap system for one-handed opening and closing. Integrated leakproof cap prevents cross contamination and lost closures. Clarified polypropylene combines chemical resistance and high speed tolerance. 15 mL and 50 mL versions are available White writing area and flat caps for sample identification. Printed graduations. Standard dimensions compatible with centrifuge rotors.

  • Hinged-cap system with ergonomic design for one-handed opening and closing
  • Integrated leakproof cap prevents cross contamination and lost closures
  • Clarified polypropylene combines chemical resistance and high speed tolerance
  • White writing area and flat caps for sample identification
  • Printed graduations
  • Standard dimensions compatible with centrifuge rotors

Legal Information

EZ Flip is a trademark of Thermo Fisher Scientific or its subsidiaries
Nunc is a registered trademark of Thermo Fisher Scientific or its subsidiaries

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sharon Maes et al.
BMC microbiology, 20(1), 373-373 (2020-12-15)
Environmental biofilms can induce attachment and protection of other microorganisms including pathogens, but can also prevent them from invasion and colonization. This opens the possibility for so-called biocontrol strategies, wherein microorganisms are applied to control the presence of other microbes.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service