AXYSCT15ML500
Centrifuge tube with screw cap, conical bottom, co-polymer
capacity 15 mL
Synonym(s):
15 ml centrifuge tubes
Select a Size
€163.00
Select a Size
About This Item
€163.00
Recommended Products
material
conical bottom tube
polypropylene
screw top cap
sterility
non-sterile
packaging
pack of 50 ea
case of 500 ea
manufacturer/tradename
Axygen Inc SCT-15ML-500
Corning
parameter
12000 × g max. RCF
capacity
15 mL
color
clear
fitting
cap (screw cap)
suitability
suitable for (centrifugation)
Looking for similar products? Visit Product Comparison Guide
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service