Key Documents
Z337277
BRAND® conical joint stopper, PP
joint: ST/NS 19/26
Select a Size
10 EA
€14.00
€14.00
Please contact Customer Service for Availability
Select a Size
Change View
10 EA
€14.00
About This Item
UNSPSC Code:
41122500
NACRES:
NB.35
€14.00
Please contact Customer Service for Availability
Joint
joint: ST/NS19/26
feature
autoclavable
manufacturer/tradename
BRAND
General description
Hollow, with sealing ribs and square grip.
Other Notes
BR144425 (India)
Legal Information
BRAND is a registered trademark of BRAND GMBH + CO KG
Choose from one of the most recent versions:
Certificates of Analysis (COA)