Key Documents
Z308668
Quickfit® conical flasks
capacity 25 mL, joint: ST/NS 14/23
Synonym(s):
FE25/1
Select a Size
Z308668-5EA
€142.00
€142.00
Please contact Customer Service for Availability
Select a Size
Change View
Z308668-5EA
€142.00
About This Item
UNSPSC Code:
41121804
NACRES:
NB.43
€142.00
Please contact Customer Service for Availability
material
borosilicate glass
conical flask
flat bottom
Joint
joint: ST/NS14/23
sterility
non-sterile
feature
closure type Standard Taper Joint
without cap
packaging
case of 5
manufacturer/tradename
Quickfit FE25/1
availability
not available in USA and Canada
capacity
25 mL
General description
With ground-glass socket for general use. Not suitable for vacuum use.
Legal Information
Quickfit is a registered trademark of SciLabware Limited
Choose from one of the most recent versions:
Certificates of Analysis (COA)