Skip to Content
Merck
All Photos(2)

Key Documents

T9772

Sigma-Aldrich

Triazolam

Synonym(s):

8-Chloro-6-[2-chlorophenyl]-1-methyl-4H-[1,2,4]triazolo[4,3-a][1,4]benzodiazepine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H12Cl2N4
CAS Number:
Molecular Weight:
343.21
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

drug control

USDEA Schedule IV; Home Office Schedule 4.1; psychotrope (France); kontrollierte Droge in Deutschland; regulated under CDSA - not available from Sigma-Aldrich Canada; psicótropo (Spain); Decreto Lei 15/93: Tabela IV (Portugal)

solubility

0.1 M HCl: soluble 1 g in 600 mL solvent
chloroform: soluble 1 g in 25 mL solvent
ethanol: soluble 1 g in 1000 mL solvent
DMSO: soluble 5.4 mg/mL
dichloromethane: 50 mg/mL, clear, colorless to faintly yellow
methanol: soluble 8.8 mg/mL
H2O: insoluble
diethyl ether: insoluble

originator

Johnson & Johnson

SMILES string

ClC1=CC=CC=C1C2=NCC3=NN=C(C)N3C4=CC=C(Cl)C=C42

InChI

1S/C17H12Cl2N4/c1-10-21-22-16-9-20-17(12-4-2-3-5-14(12)19)13-8-11(18)6-7-15(13)23(10)16/h2-8H,9H2,1H3

InChI key

JOFWLTCLBGQGBO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Triazolam has been used to define nonspecific binding in radioligand binding assays[1]. Triazolam has also been used to study the effects of reducing mPer1 mRNA levels in mouse cerbellum[2].

Biochem/physiol Actions

Anxiolytic; ligand for the GABAA receptor benzodiazepine modulatory site.

Features and Benefits

This compound is a featured product for ADME Tox research. Click here to discover more featured ADME Tox products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.
This compound was developed by Johnson & Johnson. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Preparation Note

Triazolam is soluble in dichloromethane at 50 mg/ml, with heat as needed, yielding a clear, colorless to faint yellow solution. Furthermore, 1g of triazolam is soluble in 25 mL chloroform, 1000 mL ethanol, and 600 mL 0.1 M HCl. It is also soluble in DMSO (5.4 mg/ml) and methanol (8.8 mg/ml). However, it is insoluble in water and in ether.

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Hazard Classifications

Repr. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R L Voorman et al.
The Journal of pharmacology and experimental therapeutics, 287(1), 381-388 (1998-10-09)
Administration of delavirdine, an HIV-1 reverse transcriptase inhibitor, to rats or monkeys resulted in apparent loss of hepatic microsomal CYP3A and delavirdine desalkylation activity. Human CYP3A catalyzes the formation of desalkyl delavirdine and 6'-hydroxy delavirdine, an unstable metabolite, while CYP2D6
M Akiyama et al.
British journal of pharmacology, 128(7), 1616-1622 (1999-12-22)
1. The mPer1 and mPer2 genes are putative mouse clock genes that regulate circadian oscillator present in the suprachiasmatic nucleus (SCN) neuron. While they are also expressed in the granular cell layer in the cerebellum, their function is unknown. In
R R Griffiths et al.
Neuroscience and biobehavioral reviews, 9(1), 133-151 (1985-01-01)
The abuse liability of a drug is a positive, interactive function of the reinforcing and adverse effects of the drug. The relative abuse liability of the hypnotic benzodiazepine, triazolam, has been controversial. This paper reviews animal and human studies bearing
G E Pakes et al.
Drugs, 22(2), 81-110 (1981-08-01)
Triazolam is a triazolobenzodiazepine with hypnotic properties, advocated for use in acute or chronic insomnia, situational insomnia in hospitalised patients, and insomnia associated with other disease states. As triazolam has a relatively short half-life of about 2 to 3 hours
Susan K Sullivan et al.
The Journal of pharmacology and experimental therapeutics, 311(2), 537-546 (2004-07-17)
Clinically used benzodiazepine and nonbenzodiazepine sedative-hypnotic agents for the treatment of insomnia produce their therapeutic effects through allosteric enhancement of the effects of the inhibitory neurotransmitter GABA at the GABA(A) receptor. Indiplon is a novel pyrazolopyrimidine sedative-hypnotic agent, currently in

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service