Skip to Content
Merck
All Photos(1)

Key Documents

T8784

Sigma-Aldrich

Thimerosal

BioXtra, 97-101%

Synonym(s):

2-(Ethylmercuriomercapto)benzoic acid sodium salt, Ethylmercurithiosalicylic acid sodium salt, Mercury-([o-carboxyphenyl]thio)ethyl sodium salt, Sodium ethylmercurithiosalicylate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
2-(C2H5HgS)C6H4CO2Na
CAS Number:
Molecular Weight:
404.81
Beilstein:
8169555
EC Number:
MDL number:
UNSPSC Code:
51473202
PubChem Substance ID:
NACRES:
NA.21

product line

BioXtra

Assay

97-101%

impurities

≤0.0005% Phosphorus (P)
≤0.1% Insoluble matter

mp

234-237 °C (dec.) (lit.)

cation traces

Al: ≤0.001%
Ca: ≤0.002%
Cu: ≤0.0005%
Fe: ≤0.0005%
K: ≤0.005%
Mg: ≤0.0005%
NH4+: ≤0.1%
Pb: ≤0.001%
Zn: ≤0.0005%

Mode of action

enzyme | inhibits

SMILES string

[Na+].CC[Hg]Sc1ccccc1C([O-])=O

InChI

1S/C7H6O2S.C2H5.Hg.Na/c8-7(9)5-3-1-2-4-6(5)10;1-2;;/h1-4,10H,(H,8,9);1H2,2H3;;/q;;2*+1/p-2

InChI key

RTKIYNMVFMVABJ-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

General description

Note: As part of the corporation global plan to support the Minamata Convention eliminating mercury-containing compounds, as of January 2021 customers will be required to provide an End-Use Declaration. Sales of thimerosal will be restricted for approved applications only.

Biochem/physiol Actions

Thimerosal is a sulfhydryl (thiol) oxidizing reagent capable of inhibiting a wide range of sulfhydryl-dependent enzymes and proteins such as PI3-kinase-dependent methionine synthase, glutamate transport receptors, and Na++K+ dependent ATPases.

Disclaimer

For U.S. Customers: Contains mercury; Do not place in trash - dispose according to local, state, or federal laws.

Signal Word

Danger

Hazard Classifications

Acute Tox. 1 Dermal - Acute Tox. 2 Inhalation - Acute Tox. 2 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - STOT RE 2 Oral

Target Organs

Kidney

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Soha J Chhaya et al.
Journal of neurotrauma, 36(6), 877-890 (2018-08-29)
Spinal cord injury (SCI) induces neuropathic pain that is refractory to treatment. Central and peripheral immune responses to SCI play critical roles in pain development. Although immune responses in the dorsal horn have been implicated in SCI-pain, immune mechanisms in
Pan-Chyr Yang et al.
Journal of the Formosan Medical Association = Taiwan yi zhi, 111(6), 333-339 (2012-07-04)
This study evaluated the immune response elicited by two formulations of an AS03(A)-adjuvanted H5N1 A/Indonesia/05/2005 prepandemic influenza vaccine, developed using manufacturing processes with or without thiomersal. In addition, it also evaluated compliance to the Centre for Biologics Evaluation and Research
Dorota Mrozek-Budzyn et al.
Neurotoxicology and teratology, 34(6), 592-597 (2012-10-17)
Despite the common use of Thimerosal as a preservative in childhood vaccines since the 1930s, there are not many studies on ethylmercury toxicokinetics and toxicodynamics in infants. The knowledge of ethylmercury's potential adverse effects is derived mostly from parallel methylmercury
R Chalghoumi et al.
Poultry science, 87(1), 32-40 (2007-12-15)
The present study was an attempt to raise hen egg yolk Ig (IgY) simultaneously directed against Salmonella Enteritidis (SE) and Salmonella Typhimurium (ST) in the same egg yolk. The immunopotentiating effect of 2 different adjuvants -- Freund's adjuvants (FA) and
Alberto Eugenio Tozzi et al.
Developmental medicine and child neurology, 54(9), 843-848 (2012-05-18)
The aim of this article was to explore the effect of duration of breastfeeding on neurocognitive development. The long-term effect of breastfeeding on neurodevelopment was examined through a battery of neuropsychological tests in 1403 children (693 females, 710 males; mean

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service