Skip to Content
Merck
All Photos(1)

Key Documents

PHR1587

Supelco

Thimerosal

Pharmaceutical Secondary Standard; Certified Reference Material

Synonym(s):

Thimerosal, 2-(Ethylmercuriomercapto)benzoic acid sodium salt, Ethylmercurithiosalicylic acid sodium salt, Mercury-([o-carboxyphenyl]thio)ethyl sodium salt, Sodium ethylmercurithiosalicylate

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
€228.00

€228.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
1 G
€228.00

About This Item

Linear Formula:
2-(C2H5HgS)C6H4CO2Na
CAS Number:
Molecular Weight:
404.81
Beilstein:
8169555
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

€228.00


Please contact Customer Service for Availability

Request a Bulk Order

grade

certified reference material
pharmaceutical secondary standard

Quality Level

Agency

traceable to Ph. Eur. Y0000109
traceable to USP 1659000

API family

thimerosal

CofA

current certificate can be downloaded

packaging

pkg of 1 g

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

234-237 °C (dec.) (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-30°C

SMILES string

[Na+].CC[Hg]Sc1ccccc1C([O-])=O

InChI

1S/C7H6O2S.C2H5.Hg.Na/c8-7(9)5-3-1-2-4-6(5)10;1-2;;/h1-4,10H,(H,8,9);1H2,2H3;;/q;;2*+1/p-2

InChI key

RTKIYNMVFMVABJ-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

General description

Thimerosal belongs to the group of alkyl mercury compounds. Pharmaceutical secondary standards for application in quality control provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards

Application

These Secondary Standards are qualified as Certified Reference Materials. These are suitable for use in several analytical applications including but not limited to pharma release testing, pharma method development for qualitative and quantitative analyses, food and beverage quality control testing, and other calibration requirements.
Thimerosal may be used as a pharmaceutical reference standard for the quantification of the analyte in ophthalmic solutions using spectrophotometric technique.[1]

Biochem/physiol Actions

Thimerosal is a sulfhydryl (thiol) oxidizing reagent capable of inhibiting a wide range of sulfhydryl-dependent enzymes and proteins such as PI3-kinase-dependent methionine synthase, glutamate transport receptors, and Na++K+ dependent ATPases.

Analysis Note

These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Footnote

To see an example of a Certificate of Analysis for this material enter LRAA6959 in the Documents slot below. This is an example certificate only and may not be the lot that you receive.

Disclaimer

For U.S. Customers: Contains mercury; Do not place in trash - dispose according to local, state, or federal laws.

Signal Word

Danger

Hazard Classifications

Acute Tox. 1 Dermal - Acute Tox. 2 Inhalation - Acute Tox. 2 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - STOT RE 2 Oral

Target Organs

Kidney

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Association between thimerosal-containing vaccine and autism
Hviid A, et al.
JAMA : The Journal of the American Medical Association, 290(13), 1763-1766 (2003)
Simultaneous spectrophotometric-multivariate calibration determination of several components of ophthalmic solutions: phenylephrine, chloramphenicol, antipyrine, methylparaben and thimerosal
Collado.SM, et al.
Talanta, 52(5), 909-920 (2000)
Autism Res advances (2007)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service