Skip to Content
Merck
All Photos(1)

Key Documents

T1293

Sigma-Aldrich

Nalgene® centrifuge tubes, Oak Ridge Style 3119

nominal capacity 30 mL

Synonym(s):

High-Speed ​​Centrifuge Tubes, Nalgene tubes

Sign Into View Organizational & Contract Pricing

Select a Size

10 EA
€110.00

€110.00


Please contact Customer Service for Availability


Select a Size

Change View
10 EA
€110.00

About This Item

UNSPSC Code:
41121703
NACRES:
NB.11

€110.00


Please contact Customer Service for Availability

material

polypropylene copolymer
polypropylene screw closure
round bottom tube
screw top cap (Linerless)

sterility

non-sterile

feature

RCF 50,000 × g

packaging

pack of 10 ea

manufacturer/tradename

Nalgene® 3119-0030

nominal capacity

30 mL

Looking for similar products? Visit Product Comparison Guide

General description

“Nalgene” Oak Ridge high-speed polypropylene copolymer (PPCO) centrifuge tubes demonstrate superior chemical compatibility with a broader range of laboratory reagents compared to regular polycarbonate centrifuge tubes.

Features and Benefits

  • Offers superior chemical resistance compared to polycarbonate tubes.
  • Capable of being repeatedly autoclaved, resulting in a longer lifespan than polycarbonate tubes subjected to repeated autoclaving.
  • When paired with a Nalgene sealing cap, the 30mL tubes can handle centrifugal forces up to 50,000 x g.
  • Includes polypropylene linerless closures, which are easy to clean and provide leakproof protection under normal conditions up to 10,000 x g.
  • Autoclavable and leakproof when used with the provided Nalgene closure.

Other Notes

Tubes must be filled to a minimum of 80% capacity to ensure proper performance.

Legal Information

Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service