T4418
Nalgene® centrifuge tubes, Round Style 3117
nominal capacity 38 mL
Synonym(s):
High-Speed Centrifuge Tubes, Nalgene tubes
All Photos(1)
About This Item
Recommended Products
material
polycarbonate
unlipped
sterility
non-sterile
feature
RCF 50,000 × g
packaging
pack of 10
manufacturer/tradename
Nalgene® 3117-0380
nominal capacity
38 mL
General description
These high-speed polycarbonate tubes withstand temperatures from -135° to +135°C (-211° to +275°F)
- Designed with round bottoms to fit high-speed rotors
- Molding technique produces uniform wall thickness for increased strength
- Uniform diameter (no taper) for better rotor fit and force tolerance
- Lip allows easier retrieval from rotor cavities
- Economical choice - may also be used as test tubes
- Longer-lasting than polycarbonate tubes under conditions of repeated autoclaving
- Autoclavable/Transparent
Legal Information
Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries