Skip to Content
Merck
All Photos(1)

Key Documents

T0775

Sigma-Aldrich

N,N,N-Trimethyl-4-(6-phenyl-1,3,5-hexatrien-1-yl)phenylammonium p-toluenesulfonate

≥95%

Synonym(s):

1,6-Diphenyl-1,3,5-hexatriene-4′-trimethylammonium tosylate, 1-(4-Trimethylammoniophenyl)-6-phenyl-1,3,5-hexatriene p-toluenesulfonate, 4′-(Trimethylammonio)diphenylhexatriene p-toluenesulfonate, TMA-DPH

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
€146.00

€146.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
€146.00

About This Item

Linear Formula:
C21H24N · C7H7O3S
CAS Number:
Molecular Weight:
461.62
MDL number:
UNSPSC Code:
12352211
PubChem Substance ID:
NACRES:
NA.25

€146.00


Please contact Customer Service for Availability

Request a Bulk Order

Assay

≥95%

mp

237 °C (dec.) (lit.)

fluorescence

λex 355 nm; λem ~430 nm in methanol

storage temp.

−20°C

SMILES string

Cc1ccc(cc1)S([O-])(=O)=O.C[N+](C)(C)c2ccc(\C=C\C=C\C=C\c3ccccc3)cc2

InChI

1S/C21H24N.C7H8O3S/c1-22(2,3)21-17-15-20(16-18-21)14-8-5-4-7-11-19-12-9-6-10-13-19;1-6-2-4-7(5-3-6)11(8,9)10/h4-18H,1-3H3;2-5H,1H3,(H,8,9,10)/q+1;/p-1/b5-4+,11-7+,14-8+;

InChI key

ZKARERKEBVSZCX-VMDDUYISSA-M

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Lipid marker for endocytosis.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J G Kuhry et al.
Cell biophysics, 5(2), 129-140 (1983-06-01)
Fluorescence intensity measurements and fluorescence microscopy data showed that TMA-DPH (trimethylammonium diphenylhexatriene), a cationic derivative of the fluorescence polarization probe DPH, has a considerably different behavior in L929 cultured cells than does its parent molecule. In contrast to DPH, it
D Illinger et al.
Biology of the cell, 71(3), 293-296 (1991-01-01)
The fluorescent hydrophobic plasma membrane probe, trimethylamino-diphenylhexatriene (TMA-DPH) was previously shown to follow the plasma membrane throughout its internalization and recycling process and thus to behave as a marker for endo- and exocytosis in living cell systems. In this paper
Belén Yélamos et al.
Bio-protocol, 8(23), e3108-e3108 (2018-12-05)
In order to study the mechanism underlying the Hepatitis C Virus (HCV) fusion process we have performed assays using phospholipid liposomes and a truncated form of E2 protein, E2661 (amino acids 384-661 of the HCV polyprotein) lacking the transmembrane region.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service