![N,N,N-Trimethyl-4-(6-phenyl-1,3,5-hexatrien-1-yl)phenylammonium p-toluenesulfonate ≥95%](/deepweb/assets/sigmaaldrich/product/structures/235/776/a2dc2d47-6cd7-428f-b848-b9976e9945f6/640/a2dc2d47-6cd7-428f-b848-b9976e9945f6.png)
N,N,N-Trimethyl-4-(6-phenyl-1,3,5-hexatrien-1-yl)phenylammonium p-toluenesulfonate
![N,N,N-Trimethyl-4-(6-phenyl-1,3,5-hexatrien-1-yl)phenylammonium p-toluenesulfonate ≥95%](/deepweb/assets/sigmaaldrich/product/structures/235/776/a2dc2d47-6cd7-428f-b848-b9976e9945f6/640/a2dc2d47-6cd7-428f-b848-b9976e9945f6.png)
Synonym(s):
1,6-Diphenyl-1,3,5-hexatriene-4′-trimethylammonium tosylate, 1-(4-Trimethylammoniophenyl)-6-phenyl-1,3,5-hexatriene p-toluenesulfonate, 4′-(Trimethylammonio)diphenylhexatriene p-toluenesulfonate, TMA-DPH
Linear Formula:
C21H24N · C7H7O3S
CAS Number:
Molecular Weight:
461.62
Products
Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Product Line
Purity
Quality Segment