Skip to Content
Merck
All Photos(2)

Key Documents

SRP5321

Sigma-Aldrich

EIF2AK1 (HRI), active, GST tagged human

PRECISIO®, recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution

Synonym(s):

HCR, HRI, KIAA1369

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
UNSPSC Code:
12352200
NACRES:
NA.32

biological source

human

recombinant

expressed in baculovirus infected Sf9 cells

product line

PRECISIO®

Assay

≥70% (SDS-PAGE)

form

buffered aqueous glycerol solution

specific activity

9.6-14.4 nmol/min·mg

mol wt

~120 kDa

NCBI accession no.

application(s)

cell analysis

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... EIF2AK1(27102)

General description

Eukaryotic translation initiation factor 2-α kinase 1 (EIF2AK1) or HRI is a kinase that can be inactivated by hemin. The gene encoding this protein is localized on human chromosome 7p22.1.

Biochem/physiol Actions

Eukaryotic translation initiation factor 2-α kinase 1 (EIF2AK1) modulates protein synthesis. It acts at the level of translation initiation to downregulate protein synthesis in response to stress. The protein is activated by heme deficiency and other stimuli and is a major protein kinase that phosphorylates EIF2-α. EIF2AK1 is downregulated in the majority of ovarian cancers compared with normal ovarian tissues. It functions in iron homeostasis and may play a role in hemolytic and inflammatory anemia.

Physical form

Supplied in 50 mM Tris-HCl, pH 7.5, 150 mM NaCl, 10 mM glutathione, 0.1 mM EDTA, 0.25 mM DTT, 0.1 mM PMSF, 25% glycerol.

Preparation Note

After opening, aliquot into smaller quantities and store at -70 °C. Avoid repeating handling and multiple freeze/thaw cycles.

Legal Information

PRECISIO is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Molecular characterisation of a recurrent, semi-cryptic RUNX1 translocation t(7;21) in myelodysplastic syndrome and acute myeloid leukaemia
Nicola Foster
British Journal of Haematology (2010)
HRI, a stress response eIF2a kinase, exhibits structural and functional stability at high temperature and alkaline conditions.
Bhavnani V
International Journal of Biological Macromolecules (2017)
The Host-Encoded Heme Regulated Inhibitor (HRI) Facilitates Virulence-Associated Activities of Bacterial Pathogens
Niraj Shrestha
PLoS ONE (2013)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service