Skip to Content
Merck
All Photos(2)

Key Documents

850801C

Avanti

16:0-22:6 PE

1-palmitoyl-2-docosahexaenoyl-sn-glycero-3-phosphoethanolamine, chloroform

Synonym(s):

1-hexadecanoyl-2-(4Z,7Z,10Z,13Z,16Z,19Z-docosahexaenoyl)-sn-glycero-3-phosphoethanolamine; PE(16:0/22:6(4Z,7Z,10Z,13Z,16Z,19Z))

Sign Into View Organizational & Contract Pricing

Select a Size

25 MG
€558.00

€558.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
25 MG
€558.00

About This Item

Empirical Formula (Hill Notation):
C43H74NO8P
CAS Number:
Molecular Weight:
764.02
UNSPSC Code:
51191904
NACRES:
NA.25

€558.00


Please contact Customer Service for Availability

Request a Bulk Order

Assay

>99% (TLC)

form

liquid

packaging

pkg of 1 × 2.5 mL (850801C-25mg)

manufacturer/tradename

Avanti Research - A Croda Brand 850801C

concentration

10 mg/mL (850801C-25mg)

lipid type

cardiolipins
phospholipids

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@@](COP([O-])(OCC[NH3+])=O)(OC(CC/C=C\C/C=C\C/C=C\C/C=C\C/C=C\C/C=C\CC)=O)COC(CCCCCCCCCCCCCCC)=O

InChI

1S/C43H74NO8P/c1-3-5-7-9-11-13-15-17-18-19-20-21-22-24-26-28-30-32-34-36-43(46)52-41(40-51-53(47,48)50-38-37-44)39-49-42(45)35-33-31-29-27-25-23-16-14-12-10-8-6-4-2/h5,7,11,13,17-18,20-21,24,26,30,32,41H,3-4,6,8-10,12,14-16,19,22-23,25,27-29,31,33-40,44H2

InChI key

MPWUZHVZZKSTPV-MADBQMNMSA-N

General description

16:0-22:6 PE is a major component of human spermatozoa.[1]

Application

16:0-22:6PE has been used to determine its detergent solubility with equimolar concentration of sphingomyelin/cholesterol (SM/CHOL).[2] It has also been used to study the phase behavior of mixtures of sphingomyelin.[3]

Packaging

5 mL Clear Glass Sealed Ampule (850801C-25mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 - STOT SE 3

Target Organs

Central nervous system, Liver,Kidney

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Oleic-and docosahexaenoic acid-containing phosphatidylethanolamines differentially phase separate from sphingomyelin
Shaikh S R, et al.
Biochimica et Biophysica Acta - Biomembranes, 1788(11), 2421-2426 (2009)
Analysis of the lipid composition of human and boar spermatozoa by MALDI-TOF mass spectrometry, thin layer chromatography and 31P NMR spectroscopy
Lessig J, et al.
Comparative Biochemistry and Physiology. Part B, Biochemistry & Molecular Biology, 137(2), 265-277 (2004)
Acyl chain unsaturation in PEs modulates phase separation from lipid raft molecules
Shaikh SR, et al.
Biochemical and Biophysical Research Communications, 311(3), 793-796 (2003)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service