Skip to Content
Merck
All Photos(1)

Key Documents

SRP3251

Sigma-Aldrich

SCF from rat

recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC), suitable for cell culture

Synonym(s):

Mast Cell Growth Factor (MGF), Steel Factor, Stem Cell Factor, c-Kit Ligand

Sign Into View Organizational & Contract Pricing

Select a Size

10 μG
€328.00

€328.00


Please contact Customer Service for Availability


Select a Size

Change View
10 μG
€328.00

About This Item

UNSPSC Code:
12352202
NACRES:
NA.32

€328.00


Please contact Customer Service for Availability

biological source

rat

recombinant

expressed in E. coli

Assay

≥98% (HPLC)
≥98% (SDS-PAGE)

form

lyophilized

potency

20-40 ng/mL ED50

mol wt

18.4 kDa

packaging

pkg of 10 μg

technique(s)

cell culture | mammalian: suitable

impurities

<0.1 EU/μg endotoxin, tested

color

white

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

General description

SCF is a hematopoietic growth factor that exerts its activity by signaling through the c-Kit receptor. SCF and c-Kit are essential for the survival, proliferation and differentiation of hematopoietic cells committed to the melanocyte and germ cell lineages. Human SCF manifests low activity on murine cells, while murine and rat SCF are fully active on human cells. Recombinant rat SCF is an 18.4 kDa polypeptide containing 165 amino acid residues, which corresponds to the sequence of the secreted soluble form of SCF.

Biochem/physiol Actions

SCF is a hematopoietic growth factor that exerts its activity by signaling through the c-Kit receptor. Recombinant rat SCF is an 18.4 kDa polypeptide containing 165 amino acid residues, which corresponds to the sequence of the secreted soluble form of SCF.

Sequence

MQEICRNPVT DNVKDITKLV ANLPNDYMIT LNYVAGMDVL PSHCWLRDMV THLSVSLTTL LDKFSNISEG LSNYSIIDKL GKIVDDLVAC MEENAPKNVK ESLKKPETRN FTPEEFFSIF NRSIDAFKDF MVASDTSDCV LSSTLGPEKD SRVSVTKPFM LPPVA

Physical form

Lyophilized from 10 mM Sodium Phosphate, pH 6.5.

Reconstitution

Centrifuge the vial prior to opening. Reconstitute in water to a concentration of 0.1-1.0 mg/ml. Do not vortex. This solution can be stored at 2-8°C for up to 1 week. For extended storage, it is recommended to further dilute in a buffer containing a carrier protein (example 0.1% BSA) and store in working aliquots at -20°C to -80°C.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

T R Ulich et al.
Blood, 78(3), 645-650 (1991-08-01)
Recombinant rat stem cell factor (rrSCF) administered to rats as a single intravenous injection causes a dose-dependent neutrophilia and lymphocytosis as well as the appearance of immature myeloid cells and occasional blast cells in the circulation. Neutrophilia begins at 2
D M Anderson et al.
Cell, 63(1), 235-243 (1990-10-05)
We have previously reported the identification of a novel mast cell growth factor (MGF) that was shown to be a ligand for c-kit and is encoded by a gene that maps near the steel locus on mouse chromosome 10. We
Yasuo Mori et al.
Proceedings of the National Academy of Sciences of the United States of America, 112(31), 9638-9643 (2015-07-22)
Determining the developmental pathway leading to erythrocytes and being able to isolate their progenitors are crucial to understanding and treating disorders of red cell imbalance such as anemia, myelodysplastic syndrome, and polycythemia vera. Here we show that the human erythrocyte
Sang-Goo Lee et al.
Asian-Australasian journal of animal sciences, 28(11), 1565-1572 (2015-11-19)
Porcine embryonic stem cells (pESCs) have become an advantageous experimental tool for developing therapeutic applications and producing transgenic animals. However, despite numerous reports of putative pESC lines, deriving validated pESC lines from embryos produced in vitro remains difficult. Here, we
Ramesh C Nayak et al.
The Journal of clinical investigation, 125(8), 3103-3116 (2015-07-21)
Severe congenital neutropenia (SCN) is often associated with inherited heterozygous point mutations in ELANE, which encodes neutrophil elastase (NE). However, a lack of appropriate models to recapitulate SCN has substantially hampered the understanding of the genetic etiology and pathobiology of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service