Skip to Content
Merck
All Photos(1)

Key Documents

GF312

Sigma-Aldrich

SCF Protein, Human Recombinant Animal Free

The Stem cell factor also known as ckit ligand, mast cell growth factor & steel factor (SLF), is a widely expressed 28-40kDa type I transmembrane glycoprotein. Recombinant animal free human SCF is manufactured using all non-animal reagents.

Synonym(s):

Stem Cell Factor, ckit Ligand, Mast Cell Growth Factor, Stell Factor, MGF

Sign Into View Organizational & Contract Pricing

Select a Size

10 μG
€434.00

€434.00


Estimated to ship on24 April 2025Details



Select a Size

Change View
10 μG
€434.00

About This Item

UNSPSC Code:
12352202
eCl@ss:
32160405
NACRES:
NA.77

€434.00


Estimated to ship on24 April 2025Details


recombinant

expressed in E. coli

Quality Level

Assay

>98% (SDS-PAGE and HPLC)

impurities

<0.1 μg/mg protein endotoxin (<1EU/μg)

NCBI accession no.

UniProt accession no.

shipped in

dry ice

General description

Product Source: E. coli
Stem cell factor (SCF), also known as ckit ligand (KL), mast cell growth factor (MGF), and steel factor (SLF), is a widely expressed 28-40kDa type I transmembrane glycoprotein. It promotes the survival, differentiation, and mobilization of multiple cell types including myeloid, erythroid, megakaryocytic, lymphoid, germ cell, and melanocyte progenitors. Human SCF manifests low activity on murine cells, while murine and rat SCF are fully active on human cells. Recombinant human SCF is an 18.4 kDa polypeptide containing 165 amino acid residues, which corresponds to the sequence of the secreted soluble form of SCF.

Specificity

Cross Reactivty
Monkey

Application

Research Category
Stem Cell Research
Research Sub Category
Growth Factors & Receptors

Quality

Bioactivity assay: The ED50 was determined by the dose-dependent stimulation of the proliferation of human TF-1 cells is ≤ 2.0 ng/ml, corresponding to a specific activity of ≥ 5 x 105 units/mg.

Physical form

Product is filtered through a 0.2 micron filter before lyophilization.
Recombinant animal free human SCF is manufactured using all non-animal reagents

Storage and Stability

Store at -20°C for up to 6 months from date of receipt.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service