Skip to Content
Merck
All Photos(2)

Key Documents

SRP0424

Sigma-Aldrich

Tankyrase 2 [849-1166] active human

recombinant, expressed in baculovirus infected Sf9 cells, ≥30% (SDS-PAGE)

Synonym(s):

PARP5b, PARP5c, TNKS2, Tankyrase 2, poly(ADP-ribosyl)transferase 5b

Sign Into View Organizational & Contract Pricing

Select a Size

10 μG
€648.00

€648.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
10 μG
€648.00

About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

€648.00


Please contact Customer Service for Availability

Request a Bulk Order

biological source

human

recombinant

expressed in baculovirus infected Sf9 cells

Assay

≥30% (SDS-PAGE)

form

aqueous solution

mol wt

62 kDa

packaging

pkg of 10 μg

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... TNKS2(80351)

General description

TNKS2 (TRF1-interacting ankyrin-related ADP-ribose polymerase 2), also known as PARP5B, tanykyrase-2, or TNKL (GenBank Accession No. NM_025235) amino acids 849-1166 (end) with N-terminal GST-tag, MW=62kDa, expressed in Sf9 cells via a Baculovirus expression system.

Application

Useful for the study of enzyme kinetics, screening inhibitors, and selectivity profiling.

Physical form

Formulated in 40 mM Tris-HCl, pH 8.0, 110 mM NaCl, 2.2 Mm KCl, 4 mM glutathione, 20% glycerol, and 3 mM DTT.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Natalie Sidorova et al.
Cell and tissue research, 323(1), 137-145 (2005-09-10)
Tankyrase, which functions at telomeres and other cellular compartments, is thought to be a positive regulator of telomerase; its isoenzyme tankyrase 2 has been cloned as a putative cancer antigen. This pilot immunohistochemical study was designed to examine whether tumors
P G Kaminker et al.
The Journal of biological chemistry, 276(38), 35891-35899 (2001-07-17)
Tankyrase (TANK1) is a human telomere-associated poly(ADP-ribose) polymerase (PARP) that binds the telomere-binding protein TRF1 and increases telomere length when overexpressed. Here we report characterization of a second human tankyrase, tankyrase 2 (TANK2), which can also interact with TRF1 but

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service