Skip to Content
Merck
All Photos(1)

Key Documents

SML3475

Sigma-Aldrich

PF-04965842

≥98% (HPLC)

Synonym(s):

Abrocitinib, N-[cis-3-(Methyl-7H-pyrrolo[2,3-d]pyrimidin-4-ylamino)cyclobutyl]-1-propanesulfonamide, N-{cis-3-[Methyl(7H-pyrrolo[2,3-d]pyrimidin-4-yl)amino]cyclobutyl}-propane-1-sulfonamide, PF 04965842

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
€69.90
25 MG
€282.00

€69.90


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
€69.90
25 MG
€282.00

About This Item

Empirical Formula (Hill Notation):
C14H21N5O2S
CAS Number:
Molecular Weight:
323.41
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

€69.90


Please contact Customer Service for Availability

Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

CN(C1=C2C(NC=C2)=NC=N1)[C@H]3C[C@H](C3)NS(=O)(CCC)=O

InChI

1S/C14H21N5O2S/c1-3-6-22(20,21)18-10-7-11(8-10)19(2)14-12-4-5-15-13(12)16-9-17-14/h4-5,9-11,18H,3,6-8H2,1-2H3,(H,15,16,17)/t10-,11+

InChI key

IUEWXNHSKRWHDY-PHIMTYICSA-N

Biochem/physiol Actions

Janus Kinase (JAK) inhibitor selective for JAK1 with an IC50 value of 29 nM for JAK1 compared to 803 nM for JAK2, >10000 nM for JAK3 and 1250 nM for Tyk2.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service