Skip to Content
Merck
All Photos(2)

Key Documents

SML3771

Sigma-Aldrich

LMTK3 inhibitor C28

≥98% (HPLC)

Synonym(s):

6-(4-Pyridinyl)-3-[3-(trifluoromethoxy)phenyl]imidazo[1,2-b]pyridazine, 6-(Pyridin-4-yl)-3-(3-(trifluoromethoxy)phenyl)imidazo[1,2-b]pyridazine

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
€154.00
25 MG
€656.00

€154.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
€154.00
25 MG
€656.00

About This Item

Empirical Formula (Hill Notation):
C18H11F3N4O
CAS Number:
Molecular Weight:
356.30
UNSPSC Code:
12352200
NACRES:
NA.77

€154.00


Please contact Customer Service for Availability

Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear (Warmed)

storage temp.

-10 to -25°C

SMILES string

FC(OC1=CC=CC(C2=CN=C3C=CC(C4=CC=NC=C4)=NN32)=C1)(F)F

InChI

1S/C18H11F3N4O/c19-18(20,21)26-14-3-1-2-13(10-14)16-11-23-17-5-4-15(24-25(16)17)12-6-8-22-9-7-12/h1-11H

InChI key

SFBZOJORTROAIE-UHFFFAOYSA-N

Biochem/physiol Actions

Compound C28 is an orally active, potent, ATP-competitive lemur tyrosine kinase 3 (LMTK3) inhibitor with additional inhibitory potency against only 4 other kinases among a panel of 140 (IC50 = 67 nM/LMTK3, 5 nM/CLK, 6 nM/DYRK1α, 48 nM/HIPK2, 41 nM/IRAK4). LMTK3 promotes proteasome-mediated LMTK3 degradation and decreases proliferation of cancer cultures (NCI-60 panel IC50 <10 μM) and displays antitumor efficacy in xenograft and transgenic breast cancer mouse models in vivo (10 mg/mL via dialy p.o.).
Orally active, potent, ATP-competitive lemur tyrosine kinase 3 (LMTK3), CLK, DYRK1α, HIPK2 inhibitor with anticancer efficacy in cultures and in vivo.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The Inhibitory Properties of a Novel, Selective LMTK3 Kinase Inhibitor
International Journal of Molecular Sciences, 24(1), 865-865 (2023)
LMTK3 inhibition affects microtubule stability
Molecular Cancer, 20(1), 53-53 (2021)
Angeliki Ditsiou et al.
Science advances, 6(46) (2020-11-15)
Elucidating signaling driven by lemur tyrosine kinase 3 (LMTK3) could help drug development. Here, we solve the crystal structure of LMTK3 kinase domain to 2.1Å resolution, determine its consensus motif and phosphoproteome, unveiling in vitro and in vivo LMTK3 substrates.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service