Skip to Content
Merck
All Photos(1)

Key Documents

SML3202

Sigma-Aldrich

TL02-59

≥98% (HPLC)

Synonym(s):

3-[(6,7-Dimethoxy-4-quinazolinyl)oxy]-N-[4-[(4-ethyl-1-piperazinyl)methyl]-3-(trifluoromethyl)phenyl]-4-methylbenzamide

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
€78.70
25 MG
€341.00

€78.70


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
€78.70
25 MG
€341.00

About This Item

Empirical Formula (Hill Notation):
C32H34F3N5O4
CAS Number:
Molecular Weight:
609.64
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

€78.70


Please contact Customer Service for Availability

Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

−20°C

SMILES string

O=C(NC1=CC(C(F)(F)F)=C(CN2CCN(CC)CC2)C=C1)C3=CC(OC4=C(C=C(C(OC)=C5)OC)C5=NC=N4)=C(C)C=C3

InChI

1S/C32H34F3N5O4/c1-5-39-10-12-40(13-11-39)18-22-8-9-23(15-25(22)32(33,34)35)38-30(41)21-7-6-20(2)27(14-21)44-31-24-16-28(42-3)29(43-4)17-26(24)36-19-37-31/h6-9,14-17,19H,5,10-13,18H2,1-4H3,(H,38,41)

InChI key

AMHWQBGAKJESFB-UHFFFAOYSA-N

Biochem/physiol Actions

Inhibitor of the Myeloid Src-Family Kinase Fgr
TL02-59 is a potent inhibitor of Fgr kinase, a Src-family kinase expressed in myeloid cells and associated with acute myelogenous leukemia (AML). It exhibited an IC50 value of 0.03 nM for Fgr, 0.10 nM for related Src-family kinase Lyn, with >1000-fold less potency for all other kinases tested. TL02-59 given orally in a mouse xenograft model of AML was able to eliminate spleen and peripheral blood leukemic cells and significantly reduce bone marrow engraftment.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service