SML3662
Etrasimod arginine
≥98% (HPLC)
Synonym(s):
APD 334 L-Arginine, L-Arginine mono[(3R)-7-[[4-cyclopentyl-3-(trifluoromethyl)phenyl]methoxy]-1,2,3,4-tetrahydrocyclopent[b]indol-3-yl]acetate, L-Arginine salt of (R)-2-(7-(4-cyclopentyl-3-(trifluoromethyl)benzyloxy)-1,2,3,4-tetrahydrocyclopenta[b]indol-3-yl)acetic acid
Select a Size
Select a Size
About This Item
Recommended Products
Quality Level
Assay
≥98% (HPLC)
form
powder
color
white to beige
solubility
DMSO: 2 mg/mL, clear (Warmed)
storage temp.
-10 to -25°C
SMILES string
N[C@@H](CCCNC(N)=N)C(O)=O.O=C(O)C[C@H]1CCC2=C1NC3=C2C=C(OCC4=CC=C(C5CCCC5)C(C(F)(F)F)=C4)C=C3
1 of 4
This Item | C8022 | M7033 | SML1461 |
---|---|---|---|
≥98% (HPLC) | ≥98% (TLC) | ≥98% (TLC) | ≥98% (HPLC) |
100 | - | 200 | 100 |
-10 to -25°C | −20°C | - | −20°C |
DMSO: 2 mg/mL, clear (Warmed) | methanol: 20 mg/mL, clear, colorless to faintly yellow | water: 49.00-51.00 mg/mL, clear to very slightly hazy, colorless to faintly yellow | H2O: 20 mg/mL, clear |
white to beige | - | - | white to beige |
Biochem/physiol Actions
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Need A Sample COA?
This is a sample Certificate of Analysis (COA) and may not represent a recently manufactured lot of this specific product.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service