Skip to Content
Merck
All Photos(1)

Key Documents

SML3516

Sigma-Aldrich

EN106

≥98% (HPLC)

Synonym(s):

2-Chloro-N-(2-cyanoethyl)-N-(2,3-dihydro-1,4-benzodioxin-6-yl)acetamide

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
€55.90
25 MG
€226.00

€55.90


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
€55.90
25 MG
€226.00

About This Item

Empirical Formula (Hill Notation):
C13H13ClN2O3
CAS Number:
Molecular Weight:
280.71
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

€55.90


Please contact Customer Service for Availability

Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

O=C(CCl)N(C1=CC2=C(OCCO2)C=C1)CCC#N

InChI key

GLDJSVHDOXCYPT-UHFFFAOYSA-N

Biochem/physiol Actions

Cell-permeable covalent recruiter of CUL2 E3 ligase FEM1B that binds to C186 of FEM1B
EN106 is a cell-permeable covalent recruiter of CUL2 E3 ligase FEM1B that binds to C186 of FEM1B. EN106 disrupts FEM1B interactions with the FNIP1 degron and inhibits CUL2FEM1B-dependent ubiquitylation without detectable off-target effects. A modified EN106 bifunctional analog (PROTAC) recruits FEM1B and induces targeted BRD4 and BCR-ABL/c-ABL degradation.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service