Skip to Content
Merck
All Photos(1)

Key Documents

5.05763

Sigma-Aldrich

ProRS Inhibitor, Halofuginone

Synonym(s):

ProRS Inhibitor, Halofuginone, STENOROL, HF, Hydrobromide, prolyl-tRNA synthetase Inhibitor, trans-(±)-7-Bromo-6-chloro-3-[3-(3-hydroxy-2-piperidinyl)-2-oxopropyl]-4(3H)-quinazolinone hydrobromide

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
€273.00

€273.00


Estimated to ship on02 June 2025Details


Request a Bulk Order

Select a Size

Change View
10 MG
€273.00

About This Item

Empirical Formula (Hill Notation):
C16H17BrClN3O3 · xHBr
CAS Number:
Molecular Weight:
414.68 (free base basis)
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

€273.00


Estimated to ship on02 June 2025Details


Request a Bulk Order

Assay

≥98% (HPLC)

Quality Level

form

powder

potency

18 nM IC50

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

white

solubility

DMSO: 100 mg/mL

storage temp.

2-8°C

SMILES string

[Br-].Brc1cc2c(cc1Cl)C(=O)N(C=N2)CC(=O)CC3NCCC[C@H]3O.[H+]

InChI

1S/C16H17BrClN3O3.BrH/c17-11-6-13-10(5-12(11)18)16(24)21(8-20-13)7-9(22)4-14-15(23)2-1-3-19-14;/h5-6,8,14-15,19,23H,1-4,7H2;1H/t14?,15-;/m1./s1

InChI key

SJUWEPZBTXEUMU-NUNOUFIPSA-N

General description

A cell-permeable racemic mixture of Halofuginone, a halogenated derivative of the Dichroa febriguga alkaloid Febrifugine, whose D-(+)/(2R, 3S) enantiomer serves as the active inhibitor against ProRS- (prolyl-tRNA synthetase) mediated aminoacylation by simultaneously targeting/blocking ProRS Proline-binding pocket with its hydroxypiperidine and tRNA 3′ end adenosine-binding site with its halogenated 4-quinazolinone, preventing not only ProRS-catalyzed prolyl adenylation (Pro-AMP formation), but also the subsequent Pro transfer from Pro-AMP to tRNAPro. Consistently, only proline, but not other NEAA, is able to reverse the inhibitory effect of HF in in vitro rabbit reticulocyte lysate translations (1 µM HF; 1 mM Pro) and prevent HF-induced cellular AAR (amino acid response) pathway activation (100 nM HF/2 mM Pro/MEF; 10 nM HF/1 mM Pro/Murine CD4+ CD25- T cells). Shown to inhibit bFGF-induced neovascularization in a murine corneal angiogenesis model (5 mg/kg/day via food intake) and significantly reduce the severity of myelin antigen MOG33-55-induced autoimmune EAE/encephalomyelitis in mice (2 µg/mouse/day i.p.) in vivo. Unlike AA-AMP mimetics, HF does not compete against ATP for ProRS binding. ATP, in its ProRS bound state, actually helps stabilize HF via hydrogen bond interactions.

Biochem/physiol Actions

Primary Target
prolyl-tRNA synthetase

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Zhou, H., et al. 2013, Nature494, 121.

Keller, T.L., et al. 2012, Nat. Chem. Biol.12, 311.

Sundrud, M.S., et al. 2009, Science324, 1334.
Elkin, M., et al. 2000, FASEB J.14, 2477.
Elkin, M., et al. 1999, Clin. Cancer Res.5, 1982.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Classifications

Acute Tox. 1 Dermal - Acute Tox. 2 Inhalation - Acute Tox. 2 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

6.1A - Combustible, acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service