Skip to Content
Merck
All Photos(1)

Key Documents

SML3483

Sigma-Aldrich

Dimethylaminoparthenolide

≥98% (HPLC)

Synonym(s):

(1aR,4E,7aS,8S,10aS,10bR)-8-[(Dimethylamino)methyl]-2,3,6,7,7a,8,10a,10b-octahydro-1a,5-dimethyloxireno[9,10]cyclodeca[1,2-b]furan-9(1aH)-one, DMAPT

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H27NO3
CAS Number:
Molecular Weight:
293.40
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

-10 to -25°C

SMILES string

C/C1=C\CC[C@](O2)(C)[C@@]2([H])[C@@H](OC([C@@H]3CN(C)C)=O)[C@@]3([H])CC1

InChI key

UJNSFDHVIBGEJZ-SQJADDCJSA-N

Biochem/physiol Actions

Dimethylaminoparthenolide (DMAPT) is a water-soluble analogue of parthenolide that exhibits anti-inflammatory and anti-tumor activities. It directly binds to ribosomal protein L10 (RPL10) leading to decrease the expression of either p65 or IKKγ in varies pancreatic cancer cells. Dimethylaminoparthenolide radiosensitizes non-small-cell lung carcinoma (NSCLC) by inhibition of the canonical NF-κB pathway.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

DMAPT is an Effective Radioprotector from Long-Term Radiation-Induced Damage to Normal Mouse Tissues In Vivo.
Morel KL, Ormsby RJ, Klebe S, Sweeney CJ, Sykes PJ.
Radiation Research, 192, 231-239 (2019)
NF-?B inhibition by dimethylaminoparthenolide radiosensitizes non-small-cell lung carcinoma by blocking DNA double-strand break repair.
Cell death discovery, 4, 10-10 (2018)
Cooperative down-regulation of ribosomal protein L10 and NF-?B signaling pathway is responsible for the anti-proliferative effects by DMAPT in pancreatic cancer cells.
Shi C, Wang Y, Guo Y, Chen Y, Liu N.
Oncotarget, 8, 35009-35018 (2017)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service