Skip to Content
Merck
All Photos(1)

Key Documents

324694

Sigma-Aldrich

Emodin

A cell-permeable, reversible, substrate competitive and potent p56lck tyrosine kinase inhibitor (IC₅₀ = 18.5 µM).

Synonym(s):

Emodin, 6-Methyl-1,3,8-trihydroxyanthraquinone

Sign Into View Organizational & Contract Pricing

Select a Size

50 MG
€123.00

€123.00


Estimated to ship on03 February 2025Details



Select a Size

Change View
50 MG
€123.00

About This Item

Empirical Formula (Hill Notation):
C15H10O5
CAS Number:
Molecular Weight:
270.24
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

€123.00


Estimated to ship on03 February 2025Details


Quality Level

Assay

≥90% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

orange

solubility

DMSO: 1 mg/mL
aqueous base: soluble
ethanol: soluble

shipped in

ambient

storage temp.

2-8°C

SMILES string

Oc1c2c(cc(c1)O)C(=O)c3c(c(cc(c3)C)O)C2=O

InChI

1S/C15H10O5/c1-6-2-8-12(10(17)3-6)15(20)13-9(14(8)19)4-7(16)5-11(13)18/h2-5,16-18H,1H3

InChI key

RHMXXJGYXNZAPX-UHFFFAOYSA-N

General description

A cell-permeable, reversible, substrate competitive and potent p56lck tyrosine kinase inhibitor (IC50 = 18.5 µM). Selectively blocks the growth of v-ras transformed human bronchial epithelial cells (IC50 = 15 µM). Also suppresses HER2/neu tyrosine kinase activity in HER2/neu overexpressing breast cancer cells.
A cell-permeable, reversible, substrate competitive and potent p56lck tyrosine kinase inhibitor (IC50 = 18.5 µM). Selectively blocks the growth of v-ras transformed human bronchial epithelial cells (IC50 = 14.8 µM) without affecting normal human bronchial epithelial cells, even at a concentration of 370 mM. Reported to act as an inhibitor of Tau aggregation and aid in the disassembly of PHF in vitro and in vivo.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
p56lck tyrosine kinase
Product does not compete with ATP.
Reversible: yes
Target IC50: 18.5 µM against p56lck tyrosine kinase

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution in DMSO, aliquot and freeze (-20°C). DMSO stock solutions are stable for up to 3 months at -20°C. Unstable in aqueous alkali; reconstitute just prior to use.

Other Notes

Pickhardt, M., et al. 2004. J. Biol. Chem.280, 3628.
Zhang, L., and Hung. M.C. 1996. Oncogene 12, 571.
Chan, T.C., et al. 1993. Biochem. Biophys. Res. Commun. 193, 1152.
Jayasuriya, H., et al. 1992. J. Nat. Prod.55, 696.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service