Skip to Content
Merck
All Photos(1)

Documents

SML3437

Sigma-Aldrich

Epigoitrin

≥98% (HPLC)

Synonym(s):

(+)-5-Vinyl-2-oxazolidinethione, (5R)-5-ethenyl-1,3-oxazolidine-2-thione, (R)-goitrin, CID 3032313, D-Goitrin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H7NOS
CAS Number:
Molecular Weight:
129.18
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

-10 to -25°C

Biochem/physiol Actions

Epigoitrin is a sulfur containing alkaloid isolated from Isatis indigotica (Ban Lan Gen) that exhibits antiviral, anticancer, and antithyroid activities. Epigoitrin significantly decreases the susceptibility of restraint mice to H1N1 influenza virus. Epigoitrin and goitrin appear to show virucidal effect on influenza virus A (H1N1) in vitro and in ovo.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zhuo Luo et al.
Frontiers in pharmacology, 10, 78-78 (2019-02-23)
Stress has been proven to modulate an individual's immune system through the release of pituitary and adrenal hormones such as the catecholamines, growth hormone, and glucocorticoids. These signal molecules can significantly alter the host immune system and make it susceptible
Li-Xing Nie et al.
Journal of ethnopharmacology, 251, 112550-112550 (2020-01-10)
Isatidis Radix, the sun-dried roots of Isatis indigotica Fortune ex Lindl., is one of the most usually used traditional Chinese medicines. For centuries, the herb has been employed in clinical practice for treatment of virus infection and inflammation. However, its
Lixing Nie et al.
Journal of analytical methods in chemistry, 2017, 2810565-2810565 (2017-09-13)
Radix isatidis (Banlangen) is a widely used traditional Chinese medicine for treating fever and removing toxic heat. Pharmacological studies have indicated that (R)-goitrin (epigoitrin) is one of the main constituents accounting for its antiviral activity, while (S)-goitrin (goitrin) is known

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service