Skip to Content
Merck
All Photos(1)

Key Documents

SML3573

Sigma-Aldrich

Goitrin

≥98% (HPLC)

Synonym(s):

(-)-5-Vinyl-2-oxazolidinethione, (5S)-5-Ethenyl-1,3-oxazolidine-2-thione, L-Goitrin

Sign Into View Organizational & Contract Pricing

Select a Size

25 MG
€86.00
100 MG
€347.00

€86.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
25 MG
€86.00
100 MG
€347.00

About This Item

Empirical Formula (Hill Notation):
C5H7NOS
CAS Number:
Molecular Weight:
129.18
MDL number:
UNSPSC Code:
51111800
UNSPSC Code:
12352200
NACRES:
NA.21

€86.00


Please contact Customer Service for Availability

Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

optical activity

[α]/D 85 to 67° (C=1.0g/100mL, CDCl3 )

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

S=C1NC[C@H](C=C)O1

InChI

1S/C5H7NOS/c1-2-4-3-6-5(8)7-4/h2,4H,1,3H2,(H,6,8)/t4-/m0/s1

InChI key

UZQVYLOFLQICCT-BYPYZUCNSA-N

Biochem/physiol Actions

Antiviral, anticancer, and antithyroid; show virucidal effect on influenza virus A (H1N1)
Goitrin is a sulfur containing alkaloid isolated from Isatis indigotica (Ban Lan Gen) and Brassica vegetables (yellow turnip, broccoli) that exhibits antiviral, anticancer, and antithyroid activities. Epigoitrin and goitrin appear to show virucidal effect on influenza virus A (H1N1) in vitro and in ovo.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Stereospecific Assay of (R)- and (S)-Goitrin in Commercial Formulation of Radix Isatidis by Reversed Phase High-Performance Liquid Chromatography
Lixing Nie, Zhong Dai, Shuangcheng Ma
Journal of Analytical Methods in Chemistry, 2017, 2810565-2810565 (2017)
1-5-Vinyl-2-thiooxazolidone, an antithyroid compound from yellow turnip and from Brassica seeds
The Journal of Biological Chemistry, 181(1), 121-130 (1941)
Antiviral activity of Isatidis Radix derived glucosinolate isomers and their breakdown products against influenza A in vitro/ovo and mechanism of action
Li-Xing Nie
Journal of Ethnopharmacology, 251, 112550-112550 (2020)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service