Skip to Content
Merck
All Photos(1)

Key Documents

SML1450

Sigma-Aldrich

Isobavachalcone

≥98% (HPCE)

Synonym(s):

(2E)-1-[2,4-Dihydroxy-3-(3-methyl-2-butenyl)phenyl]-3-(4-hydroxyphenyl)-2-propen-1-one, 2′,4,4′-Trihydroxy-3′-(3-methyl-2-butenyl)-chalcone, Corylifolinin, Isobacachalcone

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
€109.00
25 MG
€425.00

€109.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
€109.00
25 MG
€425.00

About This Item

Empirical Formula (Hill Notation):
C20H20O4
CAS Number:
Molecular Weight:
324.37
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

€109.00


Please contact Customer Service for Availability

Request a Bulk Order

Quality Level

Assay

≥98% (HPCE)

form

powder

storage condition

desiccated
protect from light

color

white to beige

solubility

DMSO: 5 mg/mL, clear (warmed)

storage temp.

−20°C

SMILES string

OC1=C(CC=C(C)C)C(O)=C(C(/C=C/C2=CC=C(O)C=C2)=O)C=C1

InChI

1S/C20H20O4/c1-13(2)3-9-16-19(23)12-10-17(20(16)24)18(22)11-6-14-4-7-15(21)8-5-14/h3-8,10-12,21,23-24H,9H2,1-2H3/b11-6+

InChI key

DUWPGRAKHMEPCM-IZZDOVSWSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Isobavachalcone is a chalcone isolated from multipurpose medical plant Psoralea corylifolia that inhibits LPS-induced ICAM-1 expression and leukocyte adhesion to brain endothelial cells. It appears that isobavachalcone modulates both MyD88-dependent and TRIF-dependent signaling of toll-like receptor 4 (TLR4). Isobavachalcone significantly inhibits both oligomerization and fibrillization of Aβ42. Isobavachalcone exhibits a wide spectrum of biological activities including antioxidative, antiplatelet, antimicrobial, anti-inflammatory, antitumor, and neuroprotective.
Isobavachalcone is a chalcone isolated from multipurpose medical plant Psoralea corylifolia that inhibits LPS-induced ICAM-1 expression and leukocyte adhesion to brain endothelial cells.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lishan Zhou et al.
Frontiers in pharmacology, 10, 1589-1589 (2020-03-03)
Psoralea corylifolia L. (PC) is a traditional Chinese herb used to treat yang deficiency of the spleen and kidney in pediatric disease. Our previous studies have found that PC can alleviate the liver oxidative stress of juvenile mice with nonalcoholic

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service