Skip to Content
Merck
All Photos(1)

Key Documents

SML3084

Sigma-Aldrich

Clascoterone

≥98% (HPLC)

Synonym(s):

21-Hydroxy-17-(1-oxopropoxy)pregn-4-ene-3,20-dione, C17P, CB-03-01, Cortexolone 17α-propionate

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
€78.50
25 MG
€316.00

€78.50


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
€78.50
25 MG
€316.00

About This Item

Empirical Formula (Hill Notation):
C24H34O5
CAS Number:
Molecular Weight:
402.52
MDL number:
UNSPSC Code:
51111800
NACRES:
NA.77

€78.50


Please contact Customer Service for Availability

Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

CCC(O[C@]1(C(CO)=O)CC[C@@]2([H])[C@]3([H])CCC4=CC(CC[C@]4(C)[C@@]3([H])CC[C@]12C)=O)=O

InChI

1S/C24H34O5/c1-4-21(28)29-24(20(27)14-25)12-9-19-17-6-5-15-13-16(26)7-10-22(15,2)18(17)8-11-23(19,24)3/h13,17-19,25H,4-12,14H2,1-3H3/t17-,18+,19+,22+,23+,24+/m1/s1

InChI key

GPNHMOZDMYNCPO-PDUMRIMRSA-N

Biochem/physiol Actions

Clascoterone is a high-affinity androgen receptor (AR) antagonist (antiandrogen) that antagonizes testosterone-stimulated transcription activity (in reporter cells) as well as DHT-induced production of lipids and inflammatory cytokines (in human sebocytes) in cultures. When applied topically, clascoterone is as active as cyproterone acetate, and 2-, 3-, and 4-times, respectively, more active than finasteride, flutamide, and progesterone in vivo (local antiandrogenic activity in hamster′s flank organ; 100-400 μg/50 μL acetone/animal).
High-affinity androgen receptor (AR) antagonist with antiandrogenic efficacy in vitro and in vivo.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Patrizia Ferraboschi et al.
Steroids, 128, 95-104 (2017-09-20)
Cortexolone-17α-propionate (CP) is a topically active antiandrogen useful in the treatment of skin disorders. In the solid state, three anhydrous forms of this drug (CPI, CPII and CPIII) occur, together with one hydrated crystal (CPW). The single crystal structure of
Caridad Rosette et al.
Journal of drugs in dermatology : JDD, 18(2), 197-201 (2019-02-28)
Cortexolone 17α-propionate (clascoterone) is a novel androgen antagonist that is currently being analyzed in a large phase 2 clinical trial for the topical treatment of androgenetic alopecia (AGA). While the pathogenesis of AGA is still debated, the consensus is that
Caridad Rosette et al.
Journal of drugs in dermatology : JDD, 18(5), 412-418 (2019-05-31)
Cortexolone 17α-propionate (clascoterone) is a novel topical androgen antagonist that is being analyzed for its ability to treat acne. The pathogenesis of acne is attributed to multiple factors, including altered sebum production, inflammatory processes, dysregulation of the hormone microenvironment, and
Cortexolone 17α-propionate for hidradenitis suppurativa.
Samuel Antranig Der Sarkissian et al.
Dermatologic therapy, 33(6), e14142-e14142 (2020-08-08)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service