Skip to Content
Merck
All Photos(1)

Key Documents

SML2413

Sigma-Aldrich

EU1794-27

≥98% (HPLC)

Synonym(s):

2-[[(2-Amino-4,5-dihydro-4-oxo-5-thiazolyl)acetyl]amino]-4,5,6,7-tetrahydro-1-benzo[b]thiophene-3-carboxylic acid tert-butyl ester, 2-[[(2-Imino-4-oxo-1,3-thiazolidin-5-yl)acetyl]amino]-4,5,6,7-tetrahydro-1-benzothiophene-3-carboxylic acid tert-butyl ester, EU 1794-27, Tert-butyl 2-(2-(2-imino-4-oxothiazolidin-5-yl)acetamido)-4,5,6,7-tetrahydrobenzo[b]thiophene-3-carboxylate

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
€55.80
25 MG
€225.00

€55.80


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
€55.80
25 MG
€225.00

About This Item

Empirical Formula (Hill Notation):
C18H23N3O4S2
CAS Number:
Molecular Weight:
409.52
UNSPSC Code:
12352200

€55.80


Please contact Customer Service for Availability

Request a Bulk Order

Assay

≥98% (HPLC)

form

powder

color

white to light brown

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

O=C(C(CC(NC1=C(C(OC(C)(C)C)=O)C2=C(CCCC2)S1)=O)S3)NC3=N

Biochem/physiol Actions

EU1794-27 is an N-methyl-d-aspartate (NMDA) receptor (NMDAR) positive allosteric modulator (PAM) toward GluN2A/B/C subtypes (EC50/Emax = 1.4 μM/130%/GluN2B, 2.8 μM/230%/GluN2C, 2.4 μM/250%/GluN2D with Glu/Gly = ECmax = 100/30 μM using xenopus oocytes co-expressing rat GluN1 and respective GluN2 subtype; EC50/Emax = 6.3 μM/400%/GluN2B, 5.0 μM/680%/GluN2C with Glu/Gly = 1/0.3 μM &,;2.4 μM/580%/GluN2D with Glu/Gly = 0.6/0.2 μM), while it exhibits Glu/Gly level-dependent GluN1-GluN2A-modulating activities (EC50/Emax = 7.4 μM/52%/GluN2A with Glu/Gly = 100/30 μM; 8.1 μM/340% with Glu/Gly = 2/0.6 μM).
GluN2B/C/D NMDAR positive allosteric modulator with Glu/Gly level-dependent modulating activity toward GluN2A.
NMDA receptor is important for glutamate neurotransmission. It is associated with learning ability and memory. NMDA receptor possesses two different subunits NMDAR1 and NMDAR2. Tyrosine phosphorylation controls the actions of NMDA receptors. It is also known to influence neuronal function.[1]

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

PSD-95 promotes Fyn-mediated tyrosine phosphorylation of the N-methyl-D-aspartate receptor subunit NR2A
Tezuka T, et al.
Proceedings of the National Academy of Sciences of the USA, 96(2), 435-440 (1999)
Riley Perszyk et al.
eLife, 7 (2018-05-25)
N-methyl-d-aspartate receptors (NMDARs) are an important receptor in the brain and have been implicated in multiple neurological disorders. Many non-selective NMDAR-targeting drugs are poorly tolerated, leading to efforts to target NMDAR subtypes to improve the therapeutic index. We describe here

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service