Skip to Content
Merck
All Photos(1)

Key Documents

SML1992

Sigma-Aldrich

FM-479

≥98% (HPLC)

Synonym(s):

(E)-2-Cyano-3-(5-(1-cyclohexyl-6-methyl-1,6-dihydroimidazo[4,5-d]pyrrolo[2,3-b]pyridin-2-yl)furan-2-yl)-N,N-dimethylacrylamide, 2-Cyano-3-(5-(1-cyclohexyl-6-methyl-1,6-dihydroimidazo[4,5-d]pyrrolo[2,3-b]pyridin-2-yl)furan-2-yl)-N,N-dimethylacrylamide, FM-381 Inactive Control

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
€111.00
25 MG
€426.00

€111.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
€111.00
25 MG
€426.00

About This Item

Empirical Formula (Hill Notation):
C25H26N6O2
CAS Number:
Molecular Weight:
442.51
UNSPSC Code:
12352200
NACRES:
NA.77

€111.00


Please contact Customer Service for Availability

Request a Bulk Order

Assay

≥98% (HPLC)

form

powder

color

yellow to orange

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

CN(C(/C(C#N)=C/C(O1)=CC=C1C2=NC3=CN=C(N(C)C=C4)C4=C3N2C5CCCCC5)=O)C

InChI

1S/C25H26N6O2/c1-29(2)25(32)16(14-26)13-18-9-10-21(33-18)24-28-20-15-27-23-19(11-12-30(23)3)22(20)31(24)17-7-5-4-6-8-17/h9-13,15,17H,4-8H2,1-3H3/b16-13+

InChI key

RPIGHLXKDWUGDT-DTQAZKPQSA-N

Biochem/physiol Actions

FM-479 is an FM-381 structural analog that exhibits no inhibitory potency against JAK3 or other kinases when used at the FM-381 effective dosing range of 100-300 nM. For characterization details of the active probe, FM-381, please visit the FM-381 probe summary on the Structural Genomics Consortium (SGC) website.

FM-381, the active probe, is available from Sigma. To learn more about and purchase FM-381, click here.

To learn about other SGC chemical probes, visit sigma.com/sgc
Inactive FM-381 structural analog.

Features and Benefits

FM-479 is a negative control for FM-381, a chemical probe available through a partnership with the Structural Genomics Consortium (SGC). To learn more and view other SGC chemical probes, visit sigma.com/SGC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Michael Forster et al.
Cell chemical biology, 23(11), 1335-1340 (2016-11-15)
Janus kinases (JAKs) are a family of cytoplasmatic tyrosine kinases that are attractive targets for the development of anti-inflammatory drugs given their roles in cytokine signaling. One question regarding JAKs and their inhibitors that remains under intensive debate is whether

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service