Skip to Content
Merck
All Photos(1)

Key Documents

SML1514

Sigma-Aldrich

SirReal2

≥98% (HPLC)

Synonym(s):

2-(4,6-Dimethyl-pyrimidin-2-ylsulfanyl)-N-(5-naphthalen-1-ylmethyl-thiazol-2-yl)-acetamide, SirReal 2

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
€132.00
25 MG
€354.00

€132.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
€132.00
25 MG
€354.00

About This Item

Empirical Formula (Hill Notation):
C22H20N4OS2
CAS Number:
Molecular Weight:
420.55
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

€132.00


Please contact Customer Service for Availability

Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 20 mg/mL, clear

storage temp.

−20°C

SMILES string

CC1=NC(SCC(NC2=NC=C(S2)CC3=CC=CC4=C3C=CC=C4)=O)=NC(C)=C1

InChI

1S/C22H20N4OS2/c1-14-10-15(2)25-22(24-14)28-13-20(27)26-21-23-12-18(29-21)11-17-8-5-7-16-6-3-4-9-19(16)17/h3-10,12H,11,13H2,1-2H3,(H,23,26,27)

InChI key

MENNDDDTIIZDDN-UHFFFAOYSA-N

Application

Sirtuin-rearranging ligand 2 (SirReal2) has been used as a positive control to evaluate in vivo sirtuin 2 (SIRT2) inhibition by RK-9123016[1] and NPD11033.[2]

Biochem/physiol Actions

SirReal2 is a potent and selective inhibitor of Sirt2, an NAD-dependent lysine deacetylase that has been implicated in the pathogenesis of cancer, inflammation, and neurodegeneration. SirReal2 has an IC50 value of 140 nM and is >1000-fold selective for Sirt2 over Sirt1 and Sirt3. SirReal2 is believed to act through a ligand-induced structural rearrangement of the active site, binding at an adjacent pocket to lock Sirt2 in an open conformation. SirReal2 caused to tubulin hyperacetylation in HeLa cells and destabilization of the checkpoint protein BubR1, consistent with Sirt2 inhibition.
SirReal2 is a potent and selective inhibitor of Sirt2.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Norio Kudo et al.
Philosophical transactions of the Royal Society of London. Series B, Biological sciences, 373(1748) (2018-04-25)
SIRT2 is a member of the human sirtuin family of proteins and possesses NAD+-dependent lysine deacetylase/deacylase activity. SIRT2 has been implicated in carcinogenesis in various cancers including leukaemia and is considered an attractive target for cancer therapy. Here, we identified
Asad Ali Shah et al.
Biological & pharmaceutical bulletin, 39(10), 1739-1742 (2016-10-12)
SIRT2 is a member of the human sirtuin family of proteins and possesses nicotinamide adenine dinucleotide (NAD)-dependent lysine deacetylase activity. SIRT2 has been involved in various cellular processes including gene transcription, genome constancy, and the cell cycle. In addition, SIRT2

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service