Skip to Content
Merck
All Photos(1)

Key Documents

SML1410

Sigma-Aldrich

A-366

≥98% (HPLC)

Synonym(s):

5′-Methoxy-6′-[3-(1-pyrrolidinyl)propoxy]-spiro[cyclobutane-1,3′-[3H]indol]-2′-amine

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
€128.00
25 MG
€504.00

€128.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
€128.00
25 MG
€504.00

About This Item

Empirical Formula (Hill Notation):
C19H27N3O2
CAS Number:
Molecular Weight:
329.44
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.77

€128.00


Please contact Customer Service for Availability

Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 10 mg/mL, clear

storage temp.

2-8°C

SMILES string

NC1=NC2=CC(OCCCN3CCCC3)=C(OC)C=C2C14CCC4

InChI

1S/C19H27N3O2/c1-23-16-12-14-15(21-18(20)19(14)6-4-7-19)13-17(16)24-11-5-10-22-8-2-3-9-22/h12-13H,2-11H2,1H3,(H2,20,21)

InChI key

BKCDJTRMYWSXMC-UHFFFAOYSA-N

Biochem/physiol Actions

A-366 is an SGC chemical probe for G9a/GLP, developed in collaboration with Abbvie. A-366 is a potent, selective inhibitor of the histone methyltransferase G9a. The IC50 values for G9a inhbition in enzymatic and cell based assays are 3.3 and approximately 3 μM, respectively. A-366 has little or no detectable activity against a panel of 21 other methyltransferases. For full characterization details, please visit the A-366 probe summary on the Structural Genomics Consortium (SGC) website.

To learn about other SGC chemical probes for epigenetic targets, visit sigma.com/sgc

Features and Benefits

A-366 is an epigenetic chemical probe available through a partnership with the Structural Genomics Consortium (SGC). To learn more and view other SGC epigenetic probes, visit sigma.com/SGC.
This compound is a featured product for Gene Regulation research. Click here to discover more featured Gene Regulation products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.

Other Notes

A-366 has been expertly reviewed and recommended by the Chemical Probes Portal. For more information, please visit the A-366 probe summary on the Chemical Probes Portal website.

related product

Product No.
Description
Pricing

Pictograms

Environment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

We offer a variety of small molecule research tools, such as transcription factor modulators, inhibitors of chromatin modifying enzymes, and agonists/antagonists for target identification and validation in gene regulation research; a selection of these research tools is shown below.

Questions

  1. How should stock solutions for item SML1410, A-366 be stored?

    1 answer
    1. The storage of stock solutions is recommended for 1 year at -80°C and 1 month at -20°C in solvent.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service