Skip to Content
Merck
All Photos(1)

Key Documents

5.06129

Sigma-Aldrich

α-(Methylamino)isobutyric Acid

Synonym(s):

α-(Methylamino)isobutyric Acid, Neutral Amino Acid Transporter A inhibitor, MeBIA

Sign Into View Organizational & Contract Pricing

Select a Size

250 MG
€109.00

€109.00


Estimated to ship on02 June 2025Details


Request a Bulk Order

Select a Size

Change View
250 MG
€109.00

About This Item

Empirical Formula (Hill Notation):
C5H11NO2
CAS Number:
Molecular Weight:
117.15
MDL number:
UNSPSC Code:
12352106
NACRES:
NA.77

€109.00


Estimated to ship on02 June 2025Details


Request a Bulk Order

Assay

≥98% (TLC)

Quality Level

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

white

solubility

water: 100 mM

storage temp.

−20°C

SMILES string

[N+H2](C(C)(C)C(=O)[O-])C

InChI

1S/C5H11NO2/c1-5(2,6-3)4(7)8/h6H,1-3H3,(H,7,8)

InChI key

DLAMVQGYEVKIRE-UHFFFAOYSA-N

General description

A glutamine transport inhibitor selective for Na+-dependent amino acid transport system A. Used in studying the functional roles of neurotransmissions mediated by amino acids.

Biochem/physiol Actions

Primary Target
Glutamine transporter

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Young, H., et al., 2013. Am. J. Physiology-Cell Physiology.298, C1401.

Lee, Y., et al., 2013. J. Psych. & Neurosci.: JPN.38, 183.
Blot, A., et al. 2009. Neurosci.164, 998.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service