Skip to Content
Merck
All Photos(1)

Key Documents

SML1303

Sigma-Aldrich

Olinone

≥98% (HPLC)

Synonym(s):

2,3,4,5-Tetrahydro-5-(4’-acetamidobutyl)-1H-pyrido-[4,3-b]indol-1-one

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
€118.00

€118.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
€118.00

About This Item

Empirical Formula (Hill Notation):
C17H21N3O2
CAS Number:
Molecular Weight:
299.37
UNSPSC Code:
12352200
NACRES:
NA.77

€118.00


Please contact Customer Service for Availability

Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 20 mg/mL, clear

storage temp.

−20°C

Biochem/physiol Actions

Olinone is a potent and selective inhibitor of BET (bromodomain and extraterminal domain) first bromodomain BrD1 that facilitates the progression of primary oligodendrocyte progenitors toward a differentiated phenotype.
Olinone is a potent and selective inhibitor of BET (bromodomain and extraterminal domain) first bromodomain BrD1.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service