Skip to Content
Merck
All Photos(1)

Key Documents

SML0877

Sigma-Aldrich

Gardiquimod

≥98% (HPLC)

Synonym(s):

1-(4-Amino-2-ethylaminomethylimidazo[4,5-c]quinolin-1-yl)-2-methylpropan-2-ol, 4-Amino-2-[(ethylamino)methyl]-?,?-dimethyl-1H-Imidazo[4,5-c]quinoline-1-ethanol

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
€125.00
25 MG
€488.00

€125.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
€125.00
25 MG
€488.00

About This Item

Empirical Formula (Hill Notation):
C17H23N5O
CAS Number:
Molecular Weight:
313.40
UNSPSC Code:
51111800
NACRES:
NA.77

€125.00


Please contact Customer Service for Availability

Request a Bulk Order

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 10 mg/mL, clear

storage temp.

−20°C

Biochem/physiol Actions

Gardiquimod is a potent and specific agonist for human or mouse Toll-like receptor 7 (TLR7).
Gardiquimod is a potent and specific agonist for human or mouse Toll-like receptor 7 (TLR7). Also, at high concentration gardiquimod activates TLR8. Similarly to imiquimod, gardiquimod could mimic the effects of viral nucleic acids on the immune system.
Gardiquimod is developed as a new imidazoquinoline compound. It stimulates NF-κB (nuclear factor-κB) activation in cells expressing human or murine toll like receptor 7 (TLR7).[1] Gardiquimod is found to be ten times more efficient than imiquimod. It induces splenocyte activation and prevents murine B16 melanoma growth and metastasis.[1] Gardiquimod might serve as a therapeutic agent to prevent HIV-1 (human immunodeficiency virus - 1) transmission. It hinders the life cycle of HIV-1, by blocking the action of HIV-1 reverse transcriptase.[2]

Features and Benefits

This compound is featured on the Cytokine Receptors (Interleukin-1 Receptor/TIR Family) page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The TLR7 agonists imiquimod and gardiquimod improve DC-based immunotherapy for melanoma in mice
Ma F, et al.
Cellular & Molecular Immunology, 7(5), 381-381 (2010)
Maarten Buitendijk et al.
AIDS research and human retroviruses, 29(6), 907-918 (2013-01-16)
Immune response modifiers are being studied as therapeutic agents for viral infections and cancer. These molecules include agonists for the Toll-like receptors (TLR), a family of innate immune receptors. TLR7 and 8, located in cellular endosomes, bind single-stranded RNA characteristic
Rosmely Hernandez et al.
Journal for immunotherapy of cancer, 9(9) (2021-09-04)
Immunization with tumor neoantigens is a promising vaccine approach to promote antitumor immunity due to their high immunogenicity, lack of expression in normal tissue, and preferential induction of tumor neoantigen-specific T cells, which are central mediators of the anti-cancer response.

Articles

Bioactive small molecules for immune system signaling target identification/validation and antibiotics, antivirals, and antifungals offered.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service