Skip to Content
Merck
All Photos(1)

Documents

SMB00418

Sigma-Aldrich

Cepharanthine

≥98% (HPLC)

Synonym(s):

(+)-Cepharanthine, 6′,12′-Dimethoxy-2,2′-dimethyl-6,7-(methylenebis(oxy))oxyacanthan, O-Methylcepharanoline, CEP, Cepharanthin, Cepharantin, NSC 623442, [14aS-(14aR*,26aS*)]-2,3,13,14,14a,15,26,26a-octahydro-22,30-dimethoxy-1,14-dimethyl- H-4,6:16,19-Dietheno-21,25-metheno-12H-[1,3]dioxolo[4,5-g]pyrido[2′,3′:17,18][1,10]dioxacycloeicosino[2,3,4-ij]isoquinoline

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C37H38N2O6
CAS Number:
Molecular Weight:
606.71
MDL number:
UNSPSC Code:
12352205
PubChem Substance ID:
NACRES:
NA.25

Quality Level

Assay

≥98% (HPLC)

form

powder

application(s)

metabolomics
vitamins, nutraceuticals, and natural products

storage temp.

2-8°C

SMILES string

CN1CCC2=CC3=C(OCO3)C(O4)=C2[C@@H]1CC5=CC=C(OC6=C(OC)C=CC(C[C@@H]7C8=CC4=C(OC)C=C8CCN7C)=C6)C=C5

InChI

1S/C37H38N2O6/c1-38-13-11-24-18-31(41-4)33-20-27(24)28(38)16-23-7-10-30(40-3)32(17-23)44-26-8-5-22(6-9-26)15-29-35-25(12-14-39(29)2)19-34-36(37(35)45-33)43-21-42-34/h5-10,17-20,28-29H,11-16,21H2,1-4H3/t28-,29+/m1/s1

InChI key

YVPXVXANRNDGTA-WDYNHAJCSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Cepharanthine is a biscoclaurine alkaloid found in Stephania cepharantha.

Biochem/physiol Actions

Cepharanthine has anticancer, anti-inflammatory, anti-allergic, immunomodulatory, anti-angiogenic, and apoptosis-inducing activities.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service