Skip to Content
Merck
All Photos(1)

Key Documents

SML0833

Sigma-Aldrich

HLY78

≥98% (HPLC)

Synonym(s):

4-Ethyl-5,6-Dihydro-5-methyl-[1,3]dioxolo[4,5-j]phenanthridine, 4-Ethyl-5-methyl-5,6-dihydro-[1,3]dioxolo[4,5-j]phenanthridine

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
€89.30
25 MG
€476.00

€89.30


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
€89.30
25 MG
€476.00

About This Item

Empirical Formula (Hill Notation):
C17H17NO2
CAS Number:
Molecular Weight:
267.32
UNSPSC Code:
12352200
NACRES:
NA.77

€89.30


Please contact Customer Service for Availability

Request a Bulk Order

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 5 mg/mL, clear (warmed)

storage temp.

2-8°C

Biochem/physiol Actions

Activator of the Wnt/β-Catenin signaling pathway.
HLY78 is an activator of the Wnt/β-Catenin signaling pathway. HLY78 acts upstream of β-catenin synergistically with Wnt. It is believed to bind at a cavity at the dimeric interface of the Axin DIX domain, causing a conformational change of Axin to an active state, potentiating Axin–LRP6 association and promoting Wnt signaling transduction. There are several Wnt inhibitors used to study their potential in cancer therapy but few small molecules that activate the Wnt pathway. In Zebrafish, HLY78 increased the expression of the hematopoietic stem cell (HSC) transcription factors runx1 and cmyb, showing that it might be useful in HSC expansion, which could be important in transplantation and recovery after injury.

Pictograms

Environment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service